Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20639681-20639942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PINK1
Entrez Gene ID
65018
Gene Name
PTEN induced putative kinase 1
Gene Aliases
BRPK, PARK6
UniGene
Hs.389171
Gene Chromosome Location
Chr.1: 20633455-20651511 on Build GRCh38
Transcripts
NM_032409.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3771608 c.707C>T p.T236I Chr.1: 20639923-20639923 on Build GRCh38
5537082 c.705C>T p.N235N Chr.1: 20639921-20639921 on Build GRCh38
5554962 c.681T>C p.G227G Chr.1: 20639897-20639897 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs891939804 Chr.1: 20639838-20639838 on Build GRCh38
rs751892736 Chr.1: 20639867-20639867 on Build GRCh38
rs776563194 Chr.1: 20639860-20639860 on Build GRCh38
rs45545034 Chr.1: 20639895-20639895 on Build GRCh38
rs756873821 Chr.1: 20639907-20639907 on Build GRCh38
rs944383442 Chr.1: 20639773-20639773 on Build GRCh38
rs561948589 Chr.1: 20639913-20639913 on Build GRCh38
rs367947092 Chr.1: 20639906-20639906 on Build GRCh38
rs771836537 Chr.1: 20639917-20639917 on Build GRCh38
rs144071530 Chr.1: 20639911-20639911 on Build GRCh38
rs374088758 Chr.1: 20639863-20639863 on Build GRCh38
rs1034789713 Chr.1: 20639921-20639921 on Build GRCh38
rs2275923 Chr.1: 20639819-20639819 on Build GRCh38
rs752336566 Chr.1: 20639889-20639889 on Build GRCh38
rs1017955040 Chr.1: 20639899-20639899 on Build GRCh38
rs764179131 Chr.1: 20639855-20639855 on Build GRCh38
rs2275924 Chr.1: 20639848-20639848 on Build GRCh38
rs749603470 Chr.1: 20639901-20639902 on Build GRCh38
rs753299329 Chr.1: 20639710-20639710 on Build GRCh38
rs778161479 Chr.1: 20639885-20639885 on Build GRCh38
rs138018628 Chr.1: 20639896-20639896 on Build GRCh38
rs766656906 Chr.1: 20639887-20639887 on Build GRCh38
rs1005352768 Chr.1: 20639730-20639730 on Build GRCh38
rs374218118 Chr.1: 20639854-20639854 on Build GRCh38
rs200024387 Chr.1: 20639894-20639894 on Build GRCh38
rs900236355 Chr.1: 20639793-20639793 on Build GRCh38
rs144650220 Chr.1: 20639732-20639732 on Build GRCh38
rs2275922 Chr.1: 20639815-20639815 on Build GRCh38
rs777546990 Chr.1: 20639902-20639902 on Build GRCh38
rs911798593 Chr.1: 20639772-20639772 on Build GRCh38
rs558458802 Chr.1: 20639878-20639878 on Build GRCh38
rs3819026 Chr.1: 20639749-20639749 on Build GRCh38
rs41265073 Chr.1: 20639788-20639788 on Build GRCh38
rs758714043 Chr.1: 20639882-20639882 on Build GRCh38
rs775745846 Chr.1: 20639843-20639843 on Build GRCh38

View Full Product Details