Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165991457-165991698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724058
Entrez Gene ID
102724058
Gene Name
uncharacterized LOC102724058
Gene Aliases
-
UniGene
Hs.570269
Gene Chromosome Location
Chr.2: 165957418-166036400 on Build GRCh38
Transcripts
NR_110598.1 NM_001165963.1 NM_001165964.1 NM_001202435.1 NM_006920.4 XM_011511602.2 XM_011511604.2 XM_011511605.2 XM_011511606.2 XM_017004644.1 XM_017004645.1 XM_017004646.1 XM_017004647.1 XM_017004648.1 XM_017004649.1 XM_017004650.1 XM_017004651.1 XM_017004652.1 XM_017004653.1 XM_017004654.1

Gene Symbol
SCN1A
Entrez Gene ID
6323
Gene Name
sodium voltage-gated channel alpha subunit 1
Gene Aliases
EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI
UniGene
Hs.22654
Gene Chromosome Location
Chr.2: 165989160-166149216 on Build GRCh38
Transcripts
NR_110598.1 NM_001165963.1 NM_001165964.1 NM_001202435.1 NM_006920.4 XM_011511602.2 XM_011511604.2 XM_011511605.2 XM_011511606.2 XM_017004644.1 XM_017004645.1 XM_017004646.1 XM_017004647.1 XM_017004648.1 XM_017004649.1 XM_017004650.1 XM_017004651.1 XM_017004652.1 XM_017004653.1 XM_017004654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4811170 c.5596G>T p.D1866Y Chr.2: 165991679-165991679 on Build GRCh38
4860720 c.5664G>C p.Q1888H Chr.2: 165991611-165991611 on Build GRCh38
5435017 c.5689A>G p.T1897A Chr.2: 165991553-165991553 on Build GRCh38
570810 c.5749C>T p.R1917C Chr.2: 165991493-165991493 on Build GRCh38
4812194 c.5771G>T p.R1924L Chr.2: 165991504-165991504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs794726845 Chr.2: 165991613-165991613 on Build GRCh38
rs796053104 Chr.2: 165991520-165991520 on Build GRCh38
rs121917981 Chr.2: 165991510-165991510 on Build GRCh38
rs121917956 Chr.2: 165991493-165991493 on Build GRCh38
rs121918804 Chr.2: 165991632-165991632 on Build GRCh38
rs554861427 Chr.2: 165991536-165991536 on Build GRCh38
rs779614747 Chr.2: 165991619-165991619 on Build GRCh38
rs796053087 Chr.2: 165991529-165991529 on Build GRCh38
rs796053102 Chr.2: 165991597-165991597 on Build GRCh38
rs796053103 Chr.2: 165991565-165991565 on Build GRCh38
rs796053048 Chr.2: 165991522-165991522 on Build GRCh38
rs886674709 Chr.2: 165991607-165991607 on Build GRCh38
rs148703212 Chr.2: 165991643-165991643 on Build GRCh38
rs371253263 Chr.2: 165991488-165991488 on Build GRCh38
rs121918793 Chr.2: 165991549-165991549 on Build GRCh38
rs587781149 Chr.2: 165991509-165991509 on Build GRCh38
rs376669368 Chr.2: 165991498-165991498 on Build GRCh38
rs749961557 Chr.2: 165991577-165991577 on Build GRCh38
rs1003035644 Chr.2: 165991572-165991572 on Build GRCh38
rs764501421 Chr.2: 165991567-165991567 on Build GRCh38
rs1057511899 Chr.2: 165991587-165991587 on Build GRCh38
rs748090629 Chr.2: 165991641-165991641 on Build GRCh38
rs201905405 Chr.2: 165991636-165991636 on Build GRCh38
rs796053044 Chr.2: 165991648-165991648 on Build GRCh38
rs3749029 Chr.2: 165991504-165991504 on Build GRCh38
rs1039777371 Chr.2: 165991591-165991591 on Build GRCh38
rs373896263 Chr.2: 165991540-165991540 on Build GRCh38
rs794726737 Chr.2: 165991495-165991495 on Build GRCh38
rs112244937 Chr.2: 165991639-165991639 on Build GRCh38
rs544316711 Chr.2: 165991671-165991671 on Build GRCh38
rs562208324 Chr.2: 165991594-165991594 on Build GRCh38
rs796053045 Chr.2: 165991532-165991532 on Build GRCh38
rs371741320 Chr.2: 165991571-165991571 on Build GRCh38
rs979985807 Chr.2: 165991480-165991480 on Build GRCh38
rs796053086 Chr.2: 165991563-165991564 on Build GRCh38
rs796053046 Chr.2: 165991529-165991529 on Build GRCh38
rs746651472 Chr.2: 165991620-165991620 on Build GRCh38
rs367943471 Chr.2: 165991569-165991569 on Build GRCh38
rs797044984 Chr.2: 165991527-165991531 on Build GRCh38
rs530954206 Chr.2: 165991630-165991630 on Build GRCh38
rs1057519211 Chr.2: 165991605-165991605 on Build GRCh38
rs1052689240 Chr.2: 165991554-165991554 on Build GRCh38
rs761690089 Chr.2: 165991502-165991502 on Build GRCh38
rs77216276 Chr.2: 165991541-165991541 on Build GRCh38
rs767698089 Chr.2: 165991512-165991512 on Build GRCh38
rs794726739 Chr.2: 165991601-165991601 on Build GRCh38
rs202159085 Chr.2: 165991563-165991563 on Build GRCh38
rs199988999 Chr.2: 165991505-165991505 on Build GRCh38
rs796053043 Chr.2: 165991655-165991655 on Build GRCh38
rs552999895 Chr.2: 165991623-165991623 on Build GRCh38

View Full Product Details