Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39730029-39730280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIEN1
Entrez Gene ID
84299
Gene Name
migration and invasion enhancer 1
Gene Aliases
C17orf37, C35, ORB3, RDX12, XTP4
UniGene
Hs.333526
Gene Chromosome Location
Chr.17: 39728500-39730563 on Build GRCh38
Transcripts
NM_032339.4 XM_005257736.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146680 c.178G>A p.G60R Chr.17: 39730203-39730203 on Build GRCh38
7134588 c.105C>T p.F35F Chr.17: 39730276-39730276 on Build GRCh38
6409091 c.145C>T p.Q49* Chr.17: 39730236-39730236 on Build GRCh38
6409028 c.142G>C p.E48Q Chr.17: 39730239-39730239 on Build GRCh38
5431093 c.159C>G p.I53M Chr.17: 39730222-39730222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963783466 Chr.17: 39730120-39730120 on Build GRCh38
rs750386771 Chr.17: 39730211-39730211 on Build GRCh38
rs768438104 Chr.17: 39730239-39730239 on Build GRCh38
rs768295658 Chr.17: 39730146-39730146 on Build GRCh38
rs574795547 Chr.17: 39730105-39730108 on Build GRCh38
rs759871222 Chr.17: 39730165-39730165 on Build GRCh38
rs778430406 Chr.17: 39730246-39730246 on Build GRCh38
rs368781173 Chr.17: 39730216-39730216 on Build GRCh38
rs146835988 Chr.17: 39730145-39730145 on Build GRCh38
rs940790532 Chr.17: 39730154-39730154 on Build GRCh38
rs965709326 Chr.17: 39730057-39730057 on Build GRCh38
rs938710622 Chr.17: 39730219-39730219 on Build GRCh38
rs909347960 Chr.17: 39730142-39730142 on Build GRCh38
rs35485227 Chr.17: 39730157-39730158 on Build GRCh38
rs747759967 Chr.17: 39730249-39730249 on Build GRCh38
rs765366969 Chr.17: 39730166-39730166 on Build GRCh38
rs754119605 Chr.17: 39730161-39730161 on Build GRCh38
rs758478029 Chr.17: 39730197-39730197 on Build GRCh38
rs973858451 Chr.17: 39730125-39730125 on Build GRCh38
rs555047882 Chr.17: 39730053-39730053 on Build GRCh38
rs577393231 Chr.17: 39730224-39730224 on Build GRCh38
rs771638581 Chr.17: 39730143-39730143 on Build GRCh38
rs1026803137 Chr.17: 39730138-39730138 on Build GRCh38
rs576641463 Chr.17: 39730069-39730069 on Build GRCh38
rs200527214 Chr.17: 39730199-39730199 on Build GRCh38
rs150225861 Chr.17: 39730250-39730250 on Build GRCh38
rs772826537 Chr.17: 39730253-39730253 on Build GRCh38
rs765030671 Chr.17: 39730082-39730082 on Build GRCh38
rs1055722155 Chr.17: 39730227-39730227 on Build GRCh38
rs984990357 Chr.17: 39730130-39730130 on Build GRCh38
rs766898008 Chr.17: 39730156-39730156 on Build GRCh38
rs1010941434 Chr.17: 39730073-39730073 on Build GRCh38
rs752906949 Chr.17: 39730175-39730175 on Build GRCh38
rs922202638 Chr.17: 39730083-39730083 on Build GRCh38
rs140980514 Chr.17: 39730228-39730228 on Build GRCh38
rs749919366 Chr.17: 39730090-39730090 on Build GRCh38
rs1017951870 Chr.17: 39730105-39730105 on Build GRCh38
rs1036420077 Chr.17: 39730172-39730172 on Build GRCh38
rs761215160 Chr.17: 39730147-39730147 on Build GRCh38

View Full Product Details