Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141246165-141246394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB15
Entrez Gene ID
56121
Gene Name
protocadherin beta 15
Gene Aliases
PCDH-BETA15
UniGene
Hs.130757
Gene Chromosome Location
Chr.5: 141245349-141248234 on Build GRCh38
Transcripts
NM_018935.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1434312 c.605G>A p.R202H Chr.5: 141246183-141246183 on Build GRCh38
256164 c.676G>A p.V226I Chr.5: 141246254-141246254 on Build GRCh38
5995921 c.784G>A p.V262I Chr.5: 141246362-141246362 on Build GRCh38
3661522 c.782T>C p.V261A Chr.5: 141246360-141246360 on Build GRCh38
3851599 c.664C>T p.R222* Chr.5: 141246242-141246242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981745836 Chr.5: 141246349-141246349 on Build GRCh38
rs782617420 Chr.5: 141246253-141246253 on Build GRCh38
rs200048407 Chr.5: 141246220-141246220 on Build GRCh38
rs782603524 Chr.5: 141246364-141246364 on Build GRCh38
rs782315136 Chr.5: 141246256-141246256 on Build GRCh38
rs1023251433 Chr.5: 141246337-141246337 on Build GRCh38
rs781807257 Chr.5: 141246328-141246328 on Build GRCh38
rs782565752 Chr.5: 141246326-141246326 on Build GRCh38
rs782479554 Chr.5: 141246233-141246233 on Build GRCh38
rs781882101 Chr.5: 141246295-141246295 on Build GRCh38
rs970334421 Chr.5: 141246343-141246343 on Build GRCh38
rs782336492 Chr.5: 141246282-141246282 on Build GRCh38
rs782754365 Chr.5: 141246312-141246312 on Build GRCh38
rs782159264 Chr.5: 141246298-141246298 on Build GRCh38
rs782724041 Chr.5: 141246293-141246293 on Build GRCh38
rs782735514 Chr.5: 141246234-141246234 on Build GRCh38
rs782266469 Chr.5: 141246185-141246185 on Build GRCh38
rs782718695 Chr.5: 141246219-141246219 on Build GRCh38
rs782428925 Chr.5: 141246347-141246347 on Build GRCh38
rs782503013 Chr.5: 141246314-141246314 on Build GRCh38
rs782167975 Chr.5: 141246226-141246226 on Build GRCh38
rs989315337 Chr.5: 141246371-141246371 on Build GRCh38
rs782228787 Chr.5: 141246370-141246370 on Build GRCh38
rs374929812 Chr.5: 141246345-141246345 on Build GRCh38
rs376750608 Chr.5: 141246190-141246190 on Build GRCh38
rs782291301 Chr.5: 141246246-141246246 on Build GRCh38
rs782024704 Chr.5: 141246184-141246184 on Build GRCh38
rs202000272 Chr.5: 141246239-141246239 on Build GRCh38
rs998261705 Chr.5: 141246223-141246223 on Build GRCh38
rs782498225 Chr.5: 141246252-141246252 on Build GRCh38
rs781968813 Chr.5: 141246285-141246285 on Build GRCh38
rs201493933 Chr.5: 141246329-141246329 on Build GRCh38
rs781985782 Chr.5: 141246268-141246268 on Build GRCh38
rs782133174 Chr.5: 141246280-141246280 on Build GRCh38
rs781939214 Chr.5: 141246262-141246262 on Build GRCh38
rs782659998 Chr.5: 141246245-141246245 on Build GRCh38
rs782451446 Chr.5: 141246242-141246242 on Build GRCh38
rs782057013 Chr.5: 141246216-141246216 on Build GRCh38
rs781829262 Chr.5: 141246313-141246313 on Build GRCh38
rs782777353 Chr.5: 141246227-141246227 on Build GRCh38
rs114818434 Chr.5: 141246203-141246203 on Build GRCh38

View Full Product Details