Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102851578-102851851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAH
Entrez Gene ID
5053
Gene Name
phenylalanine hydroxylase
Gene Aliases
PH, PKU, PKU1
UniGene
Hs.603740
Gene Chromosome Location
Chr.12: 102838326-102917603 on Build GRCh38
Transcripts
NM_000277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7239561 c.861G>T p.L287L Chr.12: 102851738-102851738 on Build GRCh38
3455200 c.908C>T p.S303F Chr.12: 102851691-102851691 on Build GRCh38
6135541 c.896T>A p.F299Y Chr.12: 102851703-102851703 on Build GRCh38
3455201 c.856G>A p.E286K Chr.12: 102851743-102851743 on Build GRCh38
4904541 c.865G>A p.G289R Chr.12: 102851734-102851734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032502401 Chr.12: 102851675-102851675 on Build GRCh38
rs62508739 Chr.12: 102851743-102851743 on Build GRCh38
rs5030853 Chr.12: 102851701-102851701 on Build GRCh38
rs774242938 Chr.12: 102851664-102851664 on Build GRCh38
rs199475592 Chr.12: 102851688-102851688 on Build GRCh38
rs754686336 Chr.12: 102851738-102851738 on Build GRCh38
rs973300958 Chr.12: 102851773-102851773 on Build GRCh38
rs746946875 Chr.12: 102851829-102851829 on Build GRCh38
rs762205695 Chr.12: 102851787-102851787 on Build GRCh38
rs199475660 Chr.12: 102851754-102851754 on Build GRCh38
rs919089627 Chr.12: 102851777-102851777 on Build GRCh38
rs765823928 Chr.12: 102851705-102851705 on Build GRCh38
rs796064504 Chr.12: 102851704-102851704 on Build GRCh38
rs949413902 Chr.12: 102851820-102851820 on Build GRCh38
rs62642933 Chr.12: 102851703-102851703 on Build GRCh38
rs746006793 Chr.12: 102851665-102851665 on Build GRCh38
rs62509019 Chr.12: 102851758-102851758 on Build GRCh38
rs772249197 Chr.12: 102851756-102851756 on Build GRCh38
rs281865446 Chr.12: 102851712-102851712 on Build GRCh38
rs922639027 Chr.12: 102851668-102851669 on Build GRCh38
rs281865450 Chr.12: 102851684-102851684 on Build GRCh38
rs775444121 Chr.12: 102851767-102851767 on Build GRCh38
rs556021587 Chr.12: 102851741-102851741 on Build GRCh38
rs62642920 Chr.12: 102851695-102851695 on Build GRCh38
rs894691589 Chr.12: 102851780-102851780 on Build GRCh38
rs62642939 Chr.12: 102851709-102851709 on Build GRCh38
rs765660781 Chr.12: 102851798-102851798 on Build GRCh38
rs62514956 Chr.12: 102851686-102851686 on Build GRCh38
rs751203209 Chr.12: 102851729-102851729 on Build GRCh38
rs62642919 Chr.12: 102851730-102851730 on Build GRCh38
rs370194528 Chr.12: 102851808-102851808 on Build GRCh38
rs576564915 Chr.12: 102851630-102851630 on Build GRCh38
rs768288352 Chr.12: 102851789-102851791 on Build GRCh38
rs62642945 Chr.12: 102851710-102851710 on Build GRCh38
rs62642910 Chr.12: 102851715-102851715 on Build GRCh38
rs772150451 Chr.12: 102851668-102851668 on Build GRCh38
rs781096854 Chr.12: 102851740-102851740 on Build GRCh38
rs193032469 Chr.12: 102851784-102851784 on Build GRCh38
rs768897517 Chr.12: 102851783-102851783 on Build GRCh38
rs769886088 Chr.12: 102851662-102851662 on Build GRCh38
rs199475609 Chr.12: 102851700-102851700 on Build GRCh38
rs748034432 Chr.12: 102851644-102851644 on Build GRCh38
rs62507267 Chr.12: 102851702-102851704 on Build GRCh38
rs917895089 Chr.12: 102851814-102851814 on Build GRCh38
rs199475608 Chr.12: 102851692-102851692 on Build GRCh38
rs199475582 Chr.12: 102851755-102851755 on Build GRCh38
rs773828440 Chr.12: 102851807-102851807 on Build GRCh38
rs775844366 Chr.12: 102851673-102851673 on Build GRCh38
rs781151957 Chr.12: 102851642-102851642 on Build GRCh38
rs199475583 Chr.12: 102851687-102851687 on Build GRCh38
rs281865449 Chr.12: 102851685-102851685 on Build GRCh38
rs199475682 Chr.12: 102851749-102851749 on Build GRCh38
rs930045223 Chr.12: 102851779-102851779 on Build GRCh38
rs747164559 Chr.12: 102851778-102851778 on Build GRCh38
rs199475693 Chr.12: 102851734-102851734 on Build GRCh38
rs62507327 Chr.12: 102851735-102851735 on Build GRCh38
rs62517168 Chr.12: 102851752-102851752 on Build GRCh38
rs62508693 Chr.12: 102851751-102851751 on Build GRCh38
rs886048890 Chr.12: 102851702-102851702 on Build GRCh38
rs765934604 Chr.12: 102851713-102851713 on Build GRCh38
rs199475636 Chr.12: 102851746-102851746 on Build GRCh38

View Full Product Details