Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136406932-136407205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDCCAG3
Entrez Gene ID
10807
Gene Name
serologically defined colon cancer antigen 3
Gene Aliases
NY-CO-3
UniGene
Hs.94300
Gene Chromosome Location
Chr.9: 136401922-136410602 on Build GRCh38
Transcripts
NM_001039707.1 NM_001039708.1 NM_006643.3 XM_005266050.3 XM_005266051.3 XM_011518157.2 XM_017014218.1 XM_017014219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5559053 c.790C>G p.L264V Chr.9: 136407174-136407174 on Build GRCh38
380708 c.795G>A p.R265R Chr.9: 136407169-136407169 on Build GRCh38
6879008 c.767G>A p.G256E Chr.9: 136407197-136407197 on Build GRCh38
6220687 c.773C>T p.S258F Chr.9: 136407191-136407191 on Build GRCh38
6331057 c.817G>T p.A273S Chr.9: 136407147-136407147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs15095 Chr.9: 136406987-136406987 on Build GRCh38
rs371985326 Chr.9: 136407166-136407166 on Build GRCh38
rs756290247 Chr.9: 136407101-136407101 on Build GRCh38
rs998617995 Chr.9: 136407170-136407170 on Build GRCh38
rs781126210 Chr.9: 136407171-136407171 on Build GRCh38
rs376969845 Chr.9: 136407151-136407151 on Build GRCh38
rs149424791 Chr.9: 136407103-136407103 on Build GRCh38
rs370442934 Chr.9: 136407111-136407111 on Build GRCh38
rs746983882 Chr.9: 136407160-136407160 on Build GRCh38
rs757120980 Chr.9: 136407148-136407148 on Build GRCh38
rs943841947 Chr.9: 136407127-136407127 on Build GRCh38
rs759289530 Chr.9: 136407115-136407115 on Build GRCh38
rs770898824 Chr.9: 136407112-136407112 on Build GRCh38
rs996498667 Chr.9: 136407002-136407002 on Build GRCh38
rs117348471 Chr.9: 136407039-136407039 on Build GRCh38
rs571515513 Chr.9: 136406985-136406985 on Build GRCh38
rs892648085 Chr.9: 136407174-136407174 on Build GRCh38
rs1003099807 Chr.9: 136407072-136407072 on Build GRCh38
rs746138180 Chr.9: 136407100-136407100 on Build GRCh38
rs777163936 Chr.9: 136406961-136406961 on Build GRCh38
rs1023100848 Chr.9: 136407030-136407030 on Build GRCh38
rs1014198130 Chr.9: 136407181-136407181 on Build GRCh38
rs756446930 Chr.9: 136407144-136407144 on Build GRCh38
rs3087886 Chr.9: 136407131-136407131 on Build GRCh38
rs1031176295 Chr.9: 136407085-136407085 on Build GRCh38
rs533619179 Chr.9: 136407121-136407121 on Build GRCh38
rs904321759 Chr.9: 136407052-136407052 on Build GRCh38
rs780145972 Chr.9: 136407146-136407146 on Build GRCh38
rs10706 Chr.9: 136406962-136406962 on Build GRCh38
rs752106171 Chr.9: 136407130-136407130 on Build GRCh38
rs773316163 Chr.9: 136407105-136407105 on Build GRCh38
rs999003288 Chr.9: 136407054-136407054 on Build GRCh38
rs550845442 Chr.9: 136407018-136407018 on Build GRCh38
rs1042686702 Chr.9: 136407029-136407029 on Build GRCh38
rs956008857 Chr.9: 136407090-136407090 on Build GRCh38
rs767940551 Chr.9: 136407136-136407136 on Build GRCh38
rs908392221 Chr.9: 136407126-136407126 on Build GRCh38
rs932568063 Chr.9: 136407142-136407142 on Build GRCh38
rs545251690 Chr.9: 136407089-136407089 on Build GRCh38
rs368016140 Chr.9: 136407147-136407147 on Build GRCh38
rs768115650 Chr.9: 136407175-136407176 on Build GRCh38
rs779114256 Chr.9: 136407150-136407150 on Build GRCh38
rs554400277 Chr.9: 136407098-136407098 on Build GRCh38
rs764442854 Chr.9: 136407125-136407125 on Build GRCh38
rs750744234 Chr.9: 136407140-136407140 on Build GRCh38
rs371170506 Chr.9: 136407099-136407099 on Build GRCh38
rs764808740 Chr.9: 136407117-136407117 on Build GRCh38
rs752785005 Chr.9: 136407011-136407011 on Build GRCh38
rs1013084759 Chr.9: 136407022-136407022 on Build GRCh38
rs931368260 Chr.9: 136406953-136406953 on Build GRCh38
rs776284908 Chr.9: 136407113-136407113 on Build GRCh38
rs770978543 Chr.9: 136407167-136407167 on Build GRCh38
rs766773577 Chr.9: 136406960-136406960 on Build GRCh38
rs1041338249 Chr.9: 136406974-136406974 on Build GRCh38
rs1006406590 Chr.9: 136406973-136406973 on Build GRCh38
rs866493570 Chr.9: 136407175-136407175 on Build GRCh38
rs368493343 Chr.9: 136407104-136407104 on Build GRCh38
rs1040844620 Chr.9: 136407129-136407129 on Build GRCh38

View Full Product Details