Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 88760161-88760434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKN2
Entrez Gene ID
5586
Gene Name
protein kinase N2
Gene Aliases
PAK2, PRK2, PRKCL2, PRO2042, Pak-2, STK7
UniGene
Hs.440833
Gene Chromosome Location
Chr.1: 88684144-88836255 on Build GRCh38
Transcripts
NM_001320707.1 NM_001320708.1 NM_001320709.1 NM_006256.3 XM_017001782.1 XM_017001783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6127943 c.466G>C p.E156Q Chr.1: 88760338-88760338 on Build GRCh38
1726620 c.416A>C p.K139T Chr.1: 88760288-88760288 on Build GRCh38
5053728 c.354C>T p.C118C Chr.1: 88760226-88760226 on Build GRCh38
1726622 c.435G>T p.L145F Chr.1: 88760307-88760307 on Build GRCh38
4010391 c.470A>G p.N157S Chr.1: 88760342-88760342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988476536 Chr.1: 88760233-88760233 on Build GRCh38
rs430600 Chr.1: 88760293-88760293 on Build GRCh38
rs759658716 Chr.1: 88760305-88760305 on Build GRCh38
rs768481092 Chr.1: 88760280-88760280 on Build GRCh38
rs775770433 Chr.1: 88760358-88760358 on Build GRCh38
rs749956821 Chr.1: 88760261-88760261 on Build GRCh38
rs748061161 Chr.1: 88760290-88760290 on Build GRCh38
rs202213262 Chr.1: 88760198-88760198 on Build GRCh38
rs983815402 Chr.1: 88760394-88760394 on Build GRCh38
rs774727164 Chr.1: 88760402-88760402 on Build GRCh38
rs1032206132 Chr.1: 88760210-88760210 on Build GRCh38
rs777478188 Chr.1: 88760406-88760409 on Build GRCh38
rs369178376 Chr.1: 88760281-88760281 on Build GRCh38
rs771072254 Chr.1: 88760197-88760197 on Build GRCh38
rs759786380 Chr.1: 88760203-88760203 on Build GRCh38
rs763099622 Chr.1: 88760370-88760370 on Build GRCh38
rs755666862 Chr.1: 88760263-88760263 on Build GRCh38
rs757765901 Chr.1: 88760256-88760256 on Build GRCh38
rs769831827 Chr.1: 88760331-88760331 on Build GRCh38
rs201132242 Chr.1: 88760212-88760212 on Build GRCh38
rs748955934 Chr.1: 88760269-88760269 on Build GRCh38
rs752005316 Chr.1: 88760245-88760245 on Build GRCh38
rs755966323 Chr.1: 88760392-88760392 on Build GRCh38
rs551658033 Chr.1: 88760206-88760206 on Build GRCh38
rs894421519 Chr.1: 88760347-88760347 on Build GRCh38
rs764424876 Chr.1: 88760241-88760241 on Build GRCh38
rs965419009 Chr.1: 88760243-88760243 on Build GRCh38
rs762258784 Chr.1: 88760404-88760404 on Build GRCh38
rs779723190 Chr.1: 88760267-88760267 on Build GRCh38
rs755541710 Chr.1: 88760304-88760304 on Build GRCh38
rs976482831 Chr.1: 88760279-88760279 on Build GRCh38
rs201995133 Chr.1: 88760234-88760234 on Build GRCh38
rs932430523 Chr.1: 88760393-88760393 on Build GRCh38
rs780638510 Chr.1: 88760260-88760260 on Build GRCh38
rs763373764 Chr.1: 88760227-88760227 on Build GRCh38
rs764451031 Chr.1: 88760396-88760396 on Build GRCh38

View Full Product Details