Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169705515-169705786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf120
Entrez Gene ID
387263
Gene Name
chromosome 6 open reading frame 120
Gene Aliases
-
UniGene
Hs.591375
Gene Chromosome Location
Chr.6: 169702112-169706358 on Build GRCh38
Transcripts
NM_001029863.2 NM_001317342.1 NM_018288.3 NM_133325.2 XM_017010998.1

Gene Symbol
PHF10
Entrez Gene ID
55274
Gene Name
PHD finger protein 10
Gene Aliases
BAF45A, XAP135
UniGene
Hs.435933
Gene Chromosome Location
Chr.6: 169703902-169724049 on Build GRCh38
Transcripts
NM_001029863.2 NM_001317342.1 NM_018288.3 NM_133325.2 XM_017010998.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7212975 c.1222+1G>A p.? Chr.6: 169705615-169705615 on Build GRCh38
6744906 c.1125T>C p.I375I Chr.6: 169705713-169705713 on Build GRCh38
7212976 c.958+1G>A p.? Chr.6: 169705615-169705615 on Build GRCh38
6744907 c.861T>C p.I287I Chr.6: 169705713-169705713 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369544131 Chr.6: 169705755-169705755 on Build GRCh38
rs771642444 Chr.6: 169705620-169705620 on Build GRCh38
rs763331452 Chr.6: 169705702-169705702 on Build GRCh38
rs753283883 Chr.6: 169705700-169705700 on Build GRCh38
rs994843417 Chr.6: 169705550-169705550 on Build GRCh38
rs144617367 Chr.6: 169705712-169705712 on Build GRCh38
rs759984393 Chr.6: 169705642-169705642 on Build GRCh38
rs4557522 Chr.6: 169705654-169705654 on Build GRCh38
rs979957862 Chr.6: 169705601-169705601 on Build GRCh38
rs757350691 Chr.6: 169705708-169705708 on Build GRCh38
rs772636304 Chr.6: 169705628-169705628 on Build GRCh38
rs757205358 Chr.6: 169705725-169705725 on Build GRCh38
rs770602533 Chr.6: 169705613-169705614 on Build GRCh38
rs547160384 Chr.6: 169705623-169705623 on Build GRCh38
rs772564261 Chr.6: 169705576-169705577 on Build GRCh38
rs4286788 Chr.6: 169705629-169705629 on Build GRCh38
rs757463898 Chr.6: 169705578-169705578 on Build GRCh38
rs138574457 Chr.6: 169705718-169705718 on Build GRCh38
rs776402928 Chr.6: 169705710-169705710 on Build GRCh38
rs773826145 Chr.6: 169705686-169705689 on Build GRCh38
rs373616681 Chr.6: 169705584-169705584 on Build GRCh38
rs912900520 Chr.6: 169705681-169705681 on Build GRCh38
rs200825555 Chr.6: 169705641-169705641 on Build GRCh38
rs150361706 Chr.6: 169705608-169705608 on Build GRCh38
rs748518201 Chr.6: 169705566-169705567 on Build GRCh38
rs778638360 Chr.6: 169705738-169705738 on Build GRCh38
rs926230468 Chr.6: 169705747-169705747 on Build GRCh38
rs747289621 Chr.6: 169705572-169705572 on Build GRCh38
rs373121095 Chr.6: 169705703-169705703 on Build GRCh38
rs768369809 Chr.6: 169705592-169705592 on Build GRCh38
rs747986925 Chr.6: 169705615-169705615 on Build GRCh38
rs369940135 Chr.6: 169705570-169705570 on Build GRCh38
rs371636901 Chr.6: 169705579-169705579 on Build GRCh38
rs566759957 Chr.6: 169705757-169705757 on Build GRCh38
rs760647087 Chr.6: 169705744-169705745 on Build GRCh38
rs377234354 Chr.6: 169705733-169705733 on Build GRCh38
rs766007874 Chr.6: 169705636-169705636 on Build GRCh38
rs765639666 Chr.6: 169705690-169705690 on Build GRCh38

View Full Product Details