Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101590146-101590373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCNP
Entrez Gene ID
57092
Gene Name
PEST proteolytic signal containing nuclear protein
Gene Aliases
-
UniGene
Hs.732633
Gene Chromosome Location
Chr.3: 101574094-101594465 on Build GRCh38
Transcripts
NM_001320395.1 NM_001320397.1 NM_001320398.1 NM_001320399.1 NM_001320400.1 NM_001320401.1 NM_020357.2 NR_135223.1 NR_135224.1 NR_135225.1 NR_135226.1 NR_135227.1 NR_135228.1 NR_135229.1 NR_135230.1 NR_135231.1 NR_135232.1 XM_017006922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4946454 c.355-1G>A p.? Chr.3: 101590214-101590214 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752636256 Chr.3: 101590196-101590196 on Build GRCh38
rs775632482 Chr.3: 101590316-101590316 on Build GRCh38
rs935954078 Chr.3: 101590329-101590329 on Build GRCh38
rs755401875 Chr.3: 101590286-101590286 on Build GRCh38
rs961034560 Chr.3: 101590204-101590204 on Build GRCh38
rs779237713 Chr.3: 101590296-101590296 on Build GRCh38
rs770517377 Chr.3: 101590185-101590185 on Build GRCh38
rs762127001 Chr.3: 101590286-101590287 on Build GRCh38
rs557613240 Chr.3: 101590276-101590276 on Build GRCh38
rs539436171 Chr.3: 101590279-101590279 on Build GRCh38
rs377250393 Chr.3: 101590274-101590274 on Build GRCh38
rs114009114 Chr.3: 101590212-101590212 on Build GRCh38
rs760743356 Chr.3: 101590179-101590179 on Build GRCh38
rs756870334 Chr.3: 101590223-101590223 on Build GRCh38
rs749765131 Chr.3: 101590285-101590285 on Build GRCh38
rs776932240 Chr.3: 101590212-101590213 on Build GRCh38
rs368781678 Chr.3: 101590311-101590311 on Build GRCh38
rs773446132 Chr.3: 101590305-101590305 on Build GRCh38
rs568606853 Chr.3: 101590205-101590205 on Build GRCh38
rs772191455 Chr.3: 101590301-101590301 on Build GRCh38
rs747085571 Chr.3: 101590306-101590306 on Build GRCh38
rs748488743 Chr.3: 101590297-101590297 on Build GRCh38
rs371841283 Chr.3: 101590313-101590313 on Build GRCh38
rs575568607 Chr.3: 101590277-101590277 on Build GRCh38
rs374339996 Chr.3: 101590200-101590200 on Build GRCh38
rs780829862 Chr.3: 101590263-101590263 on Build GRCh38
rs749930749 Chr.3: 101590268-101590268 on Build GRCh38
rs17851950 Chr.3: 101590237-101590237 on Build GRCh38
rs376792719 Chr.3: 101590319-101590319 on Build GRCh38
rs924103693 Chr.3: 101590184-101590184 on Build GRCh38

View Full Product Details