Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 5000562-5000787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPP40
Entrez Gene ID
10799
Gene Name
ribonuclease P/MRP subunit p40
Gene Aliases
RNASEP1, bA428J1.3
UniGene
Hs.511756
Gene Chromosome Location
Chr.6: 4992405-5004063 on Build GRCh38
Transcripts
NM_001286132.1 NM_001286133.1 NM_006638.3 XM_011514253.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5554467 c.337G>C p.G113R Chr.6: 5000563-5000563 on Build GRCh38
323125 c.307G>A p.E103K Chr.6: 5000593-5000593 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371262049 Chr.6: 5000678-5000678 on Build GRCh38
rs749808007 Chr.6: 5000725-5000725 on Build GRCh38
rs762424392 Chr.6: 5000591-5000591 on Build GRCh38
rs757565376 Chr.6: 5000652-5000652 on Build GRCh38
rs768030501 Chr.6: 5000598-5000598 on Build GRCh38
rs767491910 Chr.6: 5000646-5000647 on Build GRCh38
rs760948754 Chr.6: 5000613-5000613 on Build GRCh38
rs996508107 Chr.6: 5000689-5000689 on Build GRCh38
rs778970208 Chr.6: 5000657-5000657 on Build GRCh38
rs899474365 Chr.6: 5000635-5000635 on Build GRCh38
rs943746078 Chr.6: 5000609-5000609 on Build GRCh38
rs201110001 Chr.6: 5000616-5000616 on Build GRCh38
rs149993815 Chr.6: 5000722-5000722 on Build GRCh38
rs888692997 Chr.6: 5000709-5000709 on Build GRCh38
rs750554568 Chr.6: 5000662-5000662 on Build GRCh38
rs959726537 Chr.6: 5000669-5000669 on Build GRCh38
rs1040814112 Chr.6: 5000622-5000622 on Build GRCh38
rs993291338 Chr.6: 5000739-5000739 on Build GRCh38
rs1015678141 Chr.6: 5000715-5000715 on Build GRCh38
rs139252154 Chr.6: 5000606-5000606 on Build GRCh38
rs750732722 Chr.6: 5000682-5000682 on Build GRCh38
rs551379409 Chr.6: 5000710-5000710 on Build GRCh38
rs1026637320 Chr.6: 5000698-5000698 on Build GRCh38
rs758513013 Chr.6: 5000675-5000675 on Build GRCh38
rs754137737 Chr.6: 5000647-5000647 on Build GRCh38

View Full Product Details