Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48914917-48915178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIM2
Entrez Gene ID
11040
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
Gene Aliases
-
UniGene
Hs.727148
Gene Chromosome Location
Chr.X: 48913182-48919136 on Build GRCh38
Transcripts
NM_006875.3

SNP Detail

SNP ID Chromosome Location
rs1028077327 Chr.X: 48915014-48915014 on Build GRCh38
rs781963527 Chr.X: 48915145-48915145 on Build GRCh38
rs900636480 Chr.X: 48914968-48914968 on Build GRCh38
rs782277942 Chr.X: 48915095-48915095 on Build GRCh38
rs781895868 Chr.X: 48914979-48914979 on Build GRCh38
rs782016042 Chr.X: 48915087-48915087 on Build GRCh38
rs34905943 Chr.X: 48915069-48915070 on Build GRCh38
rs782165711 Chr.X: 48915094-48915094 on Build GRCh38
rs782498337 Chr.X: 48914999-48914999 on Build GRCh38
rs782445437 Chr.X: 48915054-48915054 on Build GRCh38
rs782239792 Chr.X: 48915033-48915033 on Build GRCh38
rs184388253 Chr.X: 48915120-48915120 on Build GRCh38
rs782048961 Chr.X: 48915146-48915146 on Build GRCh38
rs3209771 Chr.X: 48915121-48915121 on Build GRCh38
rs782388078 Chr.X: 48915029-48915029 on Build GRCh38
rs782580697 Chr.X: 48915073-48915073 on Build GRCh38
rs1015483876 Chr.X: 48915153-48915153 on Build GRCh38
rs996271105 Chr.X: 48914990-48914990 on Build GRCh38
rs952041826 Chr.X: 48915091-48915091 on Build GRCh38
rs782254272 Chr.X: 48915138-48915138 on Build GRCh38
rs1019053929 Chr.X: 48914943-48914943 on Build GRCh38
rs782264264 Chr.X: 48915127-48915127 on Build GRCh38

View Full Product Details