Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 79122774-79123037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REG1A
Entrez Gene ID
5967
Gene Name
regenerating family member 1 alpha
Gene Aliases
ICRF, P19, PSP, PSPS, PSPS1, PTP, REG
UniGene
Hs.49407
Gene Chromosome Location
Chr.2: 79120458-79123419 on Build GRCh38
Transcripts
NM_002909.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6158763 c.392G>A p.S131N Chr.2: 79122911-79122911 on Build GRCh38
1409666 c.397A>G p.N133D Chr.2: 79122916-79122916 on Build GRCh38
6158764 c.372G>T p.W124C Chr.2: 79122891-79122891 on Build GRCh38
3583538 c.338G>A p.W113* Chr.2: 79122857-79122857 on Build GRCh38
1615141 c.384C>A p.A128A Chr.2: 79122903-79122903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777419923 Chr.2: 79122887-79122887 on Build GRCh38
rs766802704 Chr.2: 79122933-79122933 on Build GRCh38
rs773624855 Chr.2: 79122912-79122912 on Build GRCh38
rs369983287 Chr.2: 79122838-79122838 on Build GRCh38
rs943362990 Chr.2: 79123015-79123015 on Build GRCh38
rs771656147 Chr.2: 79122867-79122867 on Build GRCh38
rs768822390 Chr.2: 79122829-79122829 on Build GRCh38
rs987444414 Chr.2: 79122961-79122961 on Build GRCh38
rs373247851 Chr.2: 79122879-79122879 on Build GRCh38
rs141619806 Chr.2: 79122794-79122794 on Build GRCh38
rs746769007 Chr.2: 79122888-79122888 on Build GRCh38
rs373802117 Chr.2: 79122908-79122908 on Build GRCh38
rs773643336 Chr.2: 79122845-79122845 on Build GRCh38
rs377480421 Chr.2: 79122979-79122979 on Build GRCh38
rs757147550 Chr.2: 79122890-79122890 on Build GRCh38
rs757964240 Chr.2: 79122974-79122974 on Build GRCh38
rs760036962 Chr.2: 79122941-79122941 on Build GRCh38
rs199611838 Chr.2: 79122842-79122842 on Build GRCh38
rs745683748 Chr.2: 79123004-79123004 on Build GRCh38
rs960274423 Chr.2: 79122802-79122802 on Build GRCh38
rs780991630 Chr.2: 79122992-79122992 on Build GRCh38
rs542042583 Chr.2: 79122985-79122985 on Build GRCh38
rs760942429 Chr.2: 79122917-79122917 on Build GRCh38
rs752222929 Chr.2: 79122871-79122871 on Build GRCh38
rs763736091 Chr.2: 79122977-79122977 on Build GRCh38
rs748244874 Chr.2: 79122909-79122909 on Build GRCh38
rs749327846 Chr.2: 79122819-79122819 on Build GRCh38
rs267599471 Chr.2: 79122944-79122944 on Build GRCh38
rs745868280 Chr.2: 79122905-79122905 on Build GRCh38
rs776692974 Chr.2: 79122810-79122810 on Build GRCh38
rs760092427 Chr.2: 79122952-79122953 on Build GRCh38
rs554372711 Chr.2: 79123013-79123013 on Build GRCh38
rs867438065 Chr.2: 79122939-79122939 on Build GRCh38
rs149418512 Chr.2: 79122907-79122907 on Build GRCh38
rs373908260 Chr.2: 79122834-79122842 on Build GRCh38
rs765761645 Chr.2: 79122947-79122947 on Build GRCh38
rs34209654 Chr.2: 79122936-79122936 on Build GRCh38
rs184789172 Chr.2: 79122836-79122836 on Build GRCh38
rs983728480 Chr.2: 79122866-79122866 on Build GRCh38
rs764772254 Chr.2: 79122870-79122870 on Build GRCh38
rs374824380 Chr.2: 79122847-79122847 on Build GRCh38
rs772360419 Chr.2: 79122910-79122910 on Build GRCh38
rs200685695 Chr.2: 79122848-79122848 on Build GRCh38
rs752147307 Chr.2: 79122970-79122970 on Build GRCh38
rs546109638 Chr.2: 79122855-79122855 on Build GRCh38
rs752689342 Chr.2: 79122900-79122900 on Build GRCh38
rs767884052 Chr.2: 79122844-79122844 on Build GRCh38

View Full Product Details