Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97660881-97661138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCBP1
Entrez Gene ID
4686
Gene Name
nuclear cap binding protein subunit 1
Gene Aliases
CBP80, NCBP, Sto1
UniGene
Hs.595669
Gene Chromosome Location
Chr.9: 97633423-97673748 on Build GRCh38
Transcripts
NM_002486.4 XM_017014735.1 XM_017014736.1 XM_017014737.1 XM_017014738.1 XM_006717278.1

Gene Symbol
XPA
Entrez Gene ID
7507
Gene Name
XPA, DNA damage recognition and repair factor
Gene Aliases
XP1, XPAC
UniGene
Hs.654364
Gene Chromosome Location
Chr.9: 97654398-97697409 on Build GRCh38
Transcripts
NM_002486.4 XM_017014735.1 XM_017014736.1 XM_017014737.1 XM_017014738.1 XM_006717278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6722811 c.1481C>A p.S494Y Chr.9: 97660949-97660949 on Build GRCh38
5552313 c.1597G>A p.D533N Chr.9: 97661065-97661065 on Build GRCh38
1624340 c.1545T>C p.D515D Chr.9: 97661013-97661013 on Build GRCh38
6722809 c.1600+2T>C p.? Chr.9: 97661070-97661070 on Build GRCh38
1286519 c.1538C>G p.T513S Chr.9: 97661006-97661006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768558440 Chr.9: 97661067-97661067 on Build GRCh38
rs779860066 Chr.9: 97661064-97661064 on Build GRCh38
rs767878003 Chr.9: 97660949-97660949 on Build GRCh38
rs759734280 Chr.9: 97661109-97661109 on Build GRCh38
rs774306068 Chr.9: 97661103-97661103 on Build GRCh38
rs776589336 Chr.9: 97661068-97661068 on Build GRCh38
rs375600675 Chr.9: 97660938-97660938 on Build GRCh38
rs375805053 Chr.9: 97660923-97660923 on Build GRCh38
rs112377432 Chr.9: 97660948-97660948 on Build GRCh38
rs753371425 Chr.9: 97661012-97661012 on Build GRCh38
rs771629468 Chr.9: 97661061-97661061 on Build GRCh38
rs371398928 Chr.9: 97661099-97661099 on Build GRCh38
rs768591085 Chr.9: 97660927-97660929 on Build GRCh38
rs778478202 Chr.9: 97660972-97660972 on Build GRCh38
rs755277577 Chr.9: 97660940-97660940 on Build GRCh38
rs138518441 Chr.9: 97661057-97661057 on Build GRCh38
rs771054644 Chr.9: 97661102-97661102 on Build GRCh38
rs35251482 Chr.9: 97660965-97660965 on Build GRCh38
rs756581884 Chr.9: 97660969-97660969 on Build GRCh38
rs199548018 Chr.9: 97661110-97661110 on Build GRCh38
rs756368788 Chr.9: 97660931-97660931 on Build GRCh38
rs773219039 Chr.9: 97660913-97660913 on Build GRCh38
rs748609838 Chr.9: 97661083-97661083 on Build GRCh38
rs753013996 Chr.9: 97660918-97660918 on Build GRCh38
rs756898200 Chr.9: 97661035-97661035 on Build GRCh38
rs774144155 Chr.9: 97660940-97660941 on Build GRCh38
rs368727721 Chr.9: 97661065-97661065 on Build GRCh38
rs150516377 Chr.9: 97661056-97661056 on Build GRCh38

View Full Product Details