Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 71124243-71124504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCEE
Entrez Gene ID
84693
Gene Name
methylmalonyl-CoA epimerase
Gene Aliases
GLOD2
UniGene
Hs.94949
Gene Chromosome Location
Chr.2: 71109676-71130288 on Build GRCh38
Transcripts
NM_032601.3 XM_005264613.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4701068 c.139C>T p.R47* Chr.2: 71124445-71124445 on Build GRCh38
5103608 c.207G>T p.L69L Chr.2: 71124377-71124377 on Build GRCh38
5103609 c.206T>C p.L69P Chr.2: 71124378-71124378 on Build GRCh38
5354381 c.99G>T p.Q33H Chr.2: 71124485-71124485 on Build GRCh38
4001734 c.311G>T p.R104L Chr.2: 71124273-71124273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747222856 Chr.2: 71124328-71124328 on Build GRCh38
rs761680229 Chr.2: 71124274-71124274 on Build GRCh38
rs780845647 Chr.2: 71124351-71124351 on Build GRCh38
rs776823339 Chr.2: 71124339-71124339 on Build GRCh38
rs374147386 Chr.2: 71124420-71124420 on Build GRCh38
rs940060022 Chr.2: 71124415-71124415 on Build GRCh38
rs200681141 Chr.2: 71124313-71124313 on Build GRCh38
rs745992582 Chr.2: 71124340-71124340 on Build GRCh38
rs898781563 Chr.2: 71124434-71124434 on Build GRCh38
rs368106863 Chr.2: 71124302-71124302 on Build GRCh38
rs755396214 Chr.2: 71124301-71124301 on Build GRCh38
rs779499957 Chr.2: 71124309-71124309 on Build GRCh38
rs373053776 Chr.2: 71124464-71124464 on Build GRCh38
rs745866098 Chr.2: 71124433-71124433 on Build GRCh38
rs754533473 Chr.2: 71124389-71124389 on Build GRCh38
rs768418490 Chr.2: 71124347-71124347 on Build GRCh38
rs111033538 Chr.2: 71124445-71124445 on Build GRCh38
rs755818233 Chr.2: 71124293-71124293 on Build GRCh38
rs147401037 Chr.2: 71124406-71124406 on Build GRCh38
rs758890055 Chr.2: 71124402-71124402 on Build GRCh38
rs770516763 Chr.2: 71124466-71124466 on Build GRCh38
rs778261717 Chr.2: 71124404-71124404 on Build GRCh38
rs765849421 Chr.2: 71124298-71124298 on Build GRCh38
rs762530336 Chr.2: 71124476-71124477 on Build GRCh38
rs766048433 Chr.2: 71124374-71124374 on Build GRCh38
rs771328116 Chr.2: 71124334-71124334 on Build GRCh38
rs776909625 Chr.2: 71124270-71124270 on Build GRCh38
rs765599114 Chr.2: 71124392-71124392 on Build GRCh38
rs767329950 Chr.2: 71124356-71124356 on Build GRCh38
rs772949284 Chr.2: 71124449-71124449 on Build GRCh38
rs369078225 Chr.2: 71124349-71124349 on Build GRCh38
rs749852541 Chr.2: 71124472-71124473 on Build GRCh38
rs6748672 Chr.2: 71124273-71124273 on Build GRCh38
rs778353525 Chr.2: 71124320-71124320 on Build GRCh38
rs11541017 Chr.2: 71124357-71124357 on Build GRCh38
rs146573280 Chr.2: 71124482-71124482 on Build GRCh38
rs767011431 Chr.2: 71124280-71124280 on Build GRCh38
rs142838031 Chr.2: 71124272-71124272 on Build GRCh38
rs750071752 Chr.2: 71124284-71124284 on Build GRCh38
rs377624743 Chr.2: 71124444-71124444 on Build GRCh38
rs377300377 Chr.2: 71124384-71124384 on Build GRCh38
rs187576440 Chr.2: 71124418-71124418 on Build GRCh38
rs924043923 Chr.2: 71124277-71124277 on Build GRCh38
rs772033573 Chr.2: 71124448-71124448 on Build GRCh38
rs758798866 Chr.2: 71124315-71124315 on Build GRCh38
rs199767486 Chr.2: 71124358-71124358 on Build GRCh38
rs753384165 Chr.2: 71124299-71124299 on Build GRCh38
rs369773741 Chr.2: 71124398-71124398 on Build GRCh38

View Full Product Details