Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197775668-197775915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLL
Entrez Gene ID
66037
Gene Name
boule homolog, RNA binding protein
Gene Aliases
BOULE
UniGene
Hs.169797
Gene Chromosome Location
Chr.2: 197726879-197786574 on Build GRCh38
Transcripts
NM_001284358.1 NM_001284361.1 NM_001284362.1 NM_033030.5 NM_197970.2 XM_006712715.3 XM_011511692.2 XM_011511693.2 XM_011511694.2 XM_017004773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4665947 c.339G>A p.K113K Chr.2: 197775714-197775714 on Build GRCh38
119018 c.277-2A>C p.? Chr.2: 197775742-197775742 on Build GRCh38
7204241 c.305A>T p.K102M Chr.2: 197775712-197775712 on Build GRCh38
718636 c.381G>A p.G127G Chr.2: 197775672-197775672 on Build GRCh38
3576087 c.330T>C p.Y110Y Chr.2: 197775723-197775723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748729175 Chr.2: 197775696-197775696 on Build GRCh38
rs1049194769 Chr.2: 197775885-197775885 on Build GRCh38
rs910324204 Chr.2: 197775699-197775699 on Build GRCh38
rs929550968 Chr.2: 197775883-197775883 on Build GRCh38
rs749281445 Chr.2: 197775704-197775704 on Build GRCh38
rs760733052 Chr.2: 197775786-197775786 on Build GRCh38
rs919512583 Chr.2: 197775882-197775882 on Build GRCh38
rs766194638 Chr.2: 197775790-197775790 on Build GRCh38
rs774026616 Chr.2: 197775736-197775736 on Build GRCh38
rs772978141 Chr.2: 197775758-197775758 on Build GRCh38
rs34056332 Chr.2: 197775780-197775780 on Build GRCh38
rs1001834992 Chr.2: 197775812-197775812 on Build GRCh38
rs768166327 Chr.2: 197775729-197775729 on Build GRCh38
rs904962808 Chr.2: 197775792-197775792 on Build GRCh38
rs753996571 Chr.2: 197775791-197775791 on Build GRCh38
rs577378674 Chr.2: 197775870-197775870 on Build GRCh38
rs1046595505 Chr.2: 197775764-197775764 on Build GRCh38
rs752334902 Chr.2: 197775714-197775719 on Build GRCh38
rs755832267 Chr.2: 197775748-197775751 on Build GRCh38
rs771870002 Chr.2: 197775751-197775751 on Build GRCh38
rs200336393 Chr.2: 197775739-197775739 on Build GRCh38
rs777594094 Chr.2: 197775762-197775766 on Build GRCh38
rs201559409 Chr.2: 197775764-197775765 on Build GRCh38

View Full Product Details