Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 74999659-74999888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARNMT1
Entrez Gene ID
138199
Gene Name
carnosine N-methyltransferase 1
Gene Aliases
C9orf41, UPF0586
UniGene
Hs.567688
Gene Chromosome Location
Chr.9: 74980794-75028456 on Build GRCh38
Transcripts
NM_001320497.1 NM_152420.2 NR_135282.1 XM_005251701.3 XM_017014269.1 XM_017014270.1 XM_017014271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4410753 c.592G>T p.D198Y Chr.9: 74999869-74999869 on Build GRCh38
7135425 c.676C>G p.Q226E Chr.9: 74999785-74999785 on Build GRCh38
5577299 c.707T>C p.F236S Chr.9: 74999754-74999754 on Build GRCh38
7172515 c.620C>G p.P207R Chr.9: 74999841-74999841 on Build GRCh38
3217233 c.620C>A p.P207H Chr.9: 74999841-74999841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144200089 Chr.9: 74999773-74999773 on Build GRCh38
rs753075085 Chr.9: 74999783-74999783 on Build GRCh38
rs756166868 Chr.9: 74999746-74999746 on Build GRCh38
rs1036647749 Chr.9: 74999774-74999774 on Build GRCh38
rs755167009 Chr.9: 74999694-74999694 on Build GRCh38
rs757684303 Chr.9: 74999791-74999791 on Build GRCh38
rs778301176 Chr.9: 74999762-74999762 on Build GRCh38
rs371710249 Chr.9: 74999691-74999691 on Build GRCh38
rs746179136 Chr.9: 74999801-74999801 on Build GRCh38
rs977590729 Chr.9: 74999845-74999845 on Build GRCh38
rs569770158 Chr.9: 74999803-74999803 on Build GRCh38
rs752747222 Chr.9: 74999708-74999708 on Build GRCh38
rs549867715 Chr.9: 74999703-74999703 on Build GRCh38
rs532342573 Chr.9: 74999806-74999806 on Build GRCh38
rs779233784 Chr.9: 74999798-74999798 on Build GRCh38

View Full Product Details