Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 123694554-123694811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WASL
Entrez Gene ID
8976
Gene Name
Wiskott-Aldrich syndrome like
Gene Aliases
N-WASP, NWASP, WASPB
UniGene
Hs.143728
Gene Chromosome Location
Chr.7: 123681927-123749071 on Build GRCh38
Transcripts
NM_003941.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1084951 c.784A>G p.T262A Chr.7: 123694757-123694757 on Build GRCh38
1084950 c.821G>T p.R274M Chr.7: 123694720-123694720 on Build GRCh38
6406088 c.748G>A p.E250K Chr.7: 123694793-123694793 on Build GRCh38
2767515 c.811G>A p.E271K Chr.7: 123694730-123694730 on Build GRCh38
1232499 c.806A>C p.K269T Chr.7: 123694735-123694735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369694106 Chr.7: 123694705-123694705 on Build GRCh38
rs531294650 Chr.7: 123694610-123694610 on Build GRCh38
rs755216139 Chr.7: 123694707-123694707 on Build GRCh38
rs750200995 Chr.7: 123694702-123694702 on Build GRCh38
rs775222241 Chr.7: 123694679-123694679 on Build GRCh38
rs371583425 Chr.7: 123694613-123694613 on Build GRCh38
rs919810628 Chr.7: 123694685-123694685 on Build GRCh38
rs530486148 Chr.7: 123694582-123694582 on Build GRCh38
rs958131988 Chr.7: 123694658-123694658 on Build GRCh38
rs866172215 Chr.7: 123694746-123694746 on Build GRCh38
rs397766507 Chr.7: 123694644-123694644 on Build GRCh38
rs1048109939 Chr.7: 123694780-123694780 on Build GRCh38
rs978091510 Chr.7: 123694625-123694625 on Build GRCh38
rs762304204 Chr.7: 123694682-123694682 on Build GRCh38
rs916753010 Chr.7: 123694686-123694686 on Build GRCh38
rs766647513 Chr.7: 123694704-123694704 on Build GRCh38
rs779029993 Chr.7: 123694773-123694773 on Build GRCh38
rs925351354 Chr.7: 123694634-123694634 on Build GRCh38
rs550919069 Chr.7: 123694692-123694692 on Build GRCh38
rs967105301 Chr.7: 123694605-123694605 on Build GRCh38
rs1020238808 Chr.7: 123694598-123694598 on Build GRCh38
rs35280100 Chr.7: 123694643-123694643 on Build GRCh38
rs544846187 Chr.7: 123694699-123694699 on Build GRCh38
rs769390138 Chr.7: 123694666-123694666 on Build GRCh38
rs79354451 Chr.7: 123694693-123694693 on Build GRCh38
rs943784702 Chr.7: 123694748-123694748 on Build GRCh38
rs755993650 Chr.7: 123694703-123694703 on Build GRCh38

View Full Product Details