Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15239739-15239968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
UniGene
Hs.187763
Gene Chromosome Location
Chr.19: 15236836-15332543 on Build GRCh38
Transcripts
NM_058243.2 XM_011527854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5020979 c.3290G>A p.R1097H Chr.19: 15239814-15239814 on Build GRCh38
7303838 c.3266T>C p.V1089A Chr.19: 15239926-15239926 on Build GRCh38
709707 c.3299C>T p.S1100F Chr.19: 15239805-15239805 on Build GRCh38
992126 c.3304G>A p.V1102I Chr.19: 15239800-15239800 on Build GRCh38
6912332 c.3337_3339delGAG p.E1113delE Chr.19: 15239765-15239767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187907043 Chr.19: 15239768-15239768 on Build GRCh38
rs35676845 Chr.19: 15239814-15239814 on Build GRCh38
rs372021833 Chr.19: 15239786-15239786 on Build GRCh38
rs779611413 Chr.19: 15239842-15239842 on Build GRCh38
rs747605396 Chr.19: 15239921-15239921 on Build GRCh38
rs199530457 Chr.19: 15239884-15239884 on Build GRCh38
rs776518007 Chr.19: 15239906-15239906 on Build GRCh38
rs200799413 Chr.19: 15239890-15239890 on Build GRCh38
rs371555101 Chr.19: 15239862-15239862 on Build GRCh38
rs200400609 Chr.19: 15239846-15239846 on Build GRCh38
rs200327440 Chr.19: 15239868-15239868 on Build GRCh38
rs1044584643 Chr.19: 15239772-15239772 on Build GRCh38
rs200501260 Chr.19: 15239850-15239850 on Build GRCh38
rs201082202 Chr.19: 15239840-15239840 on Build GRCh38
rs754011460 Chr.19: 15239792-15239792 on Build GRCh38
rs201646539 Chr.19: 15239803-15239803 on Build GRCh38
rs757573992 Chr.19: 15239888-15239888 on Build GRCh38
rs200882606 Chr.19: 15239785-15239785 on Build GRCh38
rs778881856 Chr.19: 15239804-15239804 on Build GRCh38
rs767605554 Chr.19: 15239780-15239780 on Build GRCh38
rs11669901 Chr.19: 15239783-15239783 on Build GRCh38
rs375354544 Chr.19: 15239828-15239828 on Build GRCh38
rs201317873 Chr.19: 15239872-15239872 on Build GRCh38
rs772125141 Chr.19: 15239858-15239858 on Build GRCh38
rs567489459 Chr.19: 15239927-15239927 on Build GRCh38
rs771140069 Chr.19: 15239774-15239776 on Build GRCh38
rs778688396 Chr.19: 15239920-15239920 on Build GRCh38
rs1046048062 Chr.19: 15239878-15239878 on Build GRCh38
rs781392567 Chr.19: 15239898-15239898 on Build GRCh38
rs201477031 Chr.19: 15239819-15239819 on Build GRCh38
rs762462881 Chr.19: 15239758-15239758 on Build GRCh38
rs201460342 Chr.19: 15239847-15239847 on Build GRCh38
rs202076913 Chr.19: 15239942-15239942 on Build GRCh38
rs773297674 Chr.19: 15239860-15239860 on Build GRCh38
rs373309106 Chr.19: 15239928-15239928 on Build GRCh38
rs547510807 Chr.19: 15239869-15239869 on Build GRCh38
rs369685357 Chr.19: 15239902-15239902 on Build GRCh38
rs995848451 Chr.19: 15239863-15239864 on Build GRCh38
rs773269235 Chr.19: 15239761-15239763 on Build GRCh38
rs920724739 Chr.19: 15239776-15239776 on Build GRCh38
rs765535087 Chr.19: 15239866-15239866 on Build GRCh38
rs747153554 Chr.19: 15239765-15239767 on Build GRCh38
rs367680758 Chr.19: 15239883-15239883 on Build GRCh38
rs757629079 Chr.19: 15239833-15239833 on Build GRCh38
rs201100073 Chr.19: 15239762-15239762 on Build GRCh38
rs201844945 Chr.19: 15239843-15239843 on Build GRCh38
rs761637834 Chr.19: 15239775-15239775 on Build GRCh38
rs778317398 Chr.19: 15239816-15239816 on Build GRCh38
rs200026456 Chr.19: 15239864-15239864 on Build GRCh38
rs775177082 Chr.19: 15239757-15239757 on Build GRCh38
rs754585456 Chr.19: 15239905-15239905 on Build GRCh38
rs752061417 Chr.19: 15239881-15239881 on Build GRCh38

View Full Product Details