Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 20077996-20078267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF90
Entrez Gene ID
7643
Gene Name
zinc finger protein 90
Gene Aliases
HTF9
UniGene
Hs.55304
Gene Chromosome Location
Chr.19: 20077994-20127032 on Build GRCh38
Transcripts
NM_007138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7241056 c.3+5G>T p.? Chr.19: 20078140-20078140 on Build GRCh38
7241057 c.3+5G>T p.? Chr.19: 20078140-20078140 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755972627 Chr.19: 20078159-20078159 on Build GRCh38
rs371269603 Chr.19: 20078043-20078043 on Build GRCh38
rs992069832 Chr.19: 20078245-20078245 on Build GRCh38
rs932800222 Chr.19: 20078050-20078050 on Build GRCh38
rs905514493 Chr.19: 20078154-20078154 on Build GRCh38
rs765385197 Chr.19: 20078116-20078116 on Build GRCh38
rs761126140 Chr.19: 20078094-20078094 on Build GRCh38
rs532790235 Chr.19: 20078221-20078221 on Build GRCh38
rs1014683653 Chr.19: 20078216-20078216 on Build GRCh38
rs919711490 Chr.19: 20078049-20078049 on Build GRCh38
rs367667711 Chr.19: 20078040-20078040 on Build GRCh38
rs759663890 Chr.19: 20078108-20078108 on Build GRCh38
rs781126976 Chr.19: 20078149-20078149 on Build GRCh38
rs1049902019 Chr.19: 20078084-20078084 on Build GRCh38
rs377037457 Chr.19: 20078199-20078199 on Build GRCh38
rs561940597 Chr.19: 20078146-20078146 on Build GRCh38
rs546232447 Chr.19: 20078036-20078036 on Build GRCh38
rs1002016419 Chr.19: 20078230-20078230 on Build GRCh38
rs951847841 Chr.19: 20078198-20078198 on Build GRCh38
rs573066169 Chr.19: 20078125-20078125 on Build GRCh38
rs749030680 Chr.19: 20078173-20078173 on Build GRCh38
rs970182801 Chr.19: 20078223-20078223 on Build GRCh38
rs749083313 Chr.19: 20078019-20078019 on Build GRCh38
rs939737925 Chr.19: 20078034-20078034 on Build GRCh38
rs1023916789 Chr.19: 20078147-20078147 on Build GRCh38
rs1041131628 Chr.19: 20078024-20078024 on Build GRCh38
rs191118346 Chr.19: 20078142-20078142 on Build GRCh38
rs747357986 Chr.19: 20078031-20078031 on Build GRCh38
rs544194118 Chr.19: 20078187-20078187 on Build GRCh38
rs772083454 Chr.19: 20078121-20078121 on Build GRCh38
rs760862756 Chr.19: 20078207-20078207 on Build GRCh38
rs1033518022 Chr.19: 20078231-20078232 on Build GRCh38
rs754088212 Chr.19: 20078020-20078020 on Build GRCh38
rs768145527 Chr.19: 20078090-20078090 on Build GRCh38
rs773816931 Chr.19: 20078092-20078092 on Build GRCh38
rs766630559 Chr.19: 20078102-20078102 on Build GRCh38
rs768934856 Chr.19: 20078238-20078238 on Build GRCh38
rs768211690 Chr.19: 20078174-20078174 on Build GRCh38
rs575529892 Chr.19: 20078162-20078162 on Build GRCh38
rs186662638 Chr.19: 20078026-20078026 on Build GRCh38
rs754131237 Chr.19: 20078106-20078106 on Build GRCh38
rs201415273 Chr.19: 20078189-20078189 on Build GRCh38
rs891786165 Chr.19: 20078171-20078171 on Build GRCh38
rs750304184 Chr.19: 20078153-20078153 on Build GRCh38
rs778644094 Chr.19: 20078184-20078184 on Build GRCh38
rs761352066 Chr.19: 20078088-20078088 on Build GRCh38
rs114144553 Chr.19: 20078120-20078120 on Build GRCh38

View Full Product Details