Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 102868934-102869171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISD2
Entrez Gene ID
493856
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
ERIS, Miner1, NAF-1, WFS2, ZCD2
UniGene
Hs.444955
Gene Chromosome Location
Chr.4: 102868978-102892807 on Build GRCh38
Transcripts
NM_001008388.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5026194 c.54G>A p.L18L Chr.4: 102869138-102869138 on Build GRCh38
2951974 c.47C>T p.A16V Chr.4: 102869131-102869131 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs572196276 Chr.4: 102868996-102868996 on Build GRCh38
rs752651575 Chr.4: 102869094-102869094 on Build GRCh38
rs1055484125 Chr.4: 102869105-102869105 on Build GRCh38
rs532346518 Chr.4: 102868966-102868966 on Build GRCh38
rs772403549 Chr.4: 102869150-102869150 on Build GRCh38
rs551895041 Chr.4: 102869056-102869056 on Build GRCh38
rs564095499 Chr.4: 102869144-102869144 on Build GRCh38
rs1052097472 Chr.4: 102869028-102869028 on Build GRCh38
rs145312923 Chr.4: 102869096-102869096 on Build GRCh38
rs922492618 Chr.4: 102868983-102868983 on Build GRCh38
rs142339135 Chr.4: 102869136-102869136 on Build GRCh38
rs985112678 Chr.4: 102868989-102868989 on Build GRCh38
rs925352549 Chr.4: 102868986-102868986 on Build GRCh38
rs774974208 Chr.4: 102869061-102869061 on Build GRCh38
rs751241670 Chr.4: 102869099-102869099 on Build GRCh38
rs1006720004 Chr.4: 102869088-102869088 on Build GRCh38
rs773366048 Chr.4: 102869073-102869073 on Build GRCh38
rs374512796 Chr.4: 102869132-102869132 on Build GRCh38
rs777634796 Chr.4: 102869098-102869098 on Build GRCh38
rs1012487113 Chr.4: 102869058-102869058 on Build GRCh38
rs762124130 Chr.4: 102869063-102869063 on Build GRCh38
rs527237189 Chr.4: 102869038-102869038 on Build GRCh38
rs223332 Chr.4: 102869029-102869029 on Build GRCh38
rs770429715 Chr.4: 102869051-102869051 on Build GRCh38
rs746657323 Chr.4: 102869042-102869042 on Build GRCh38
rs34403962 Chr.4: 102869044-102869044 on Build GRCh38
rs571899037 Chr.4: 102869067-102869067 on Build GRCh38
rs374131019 Chr.4: 102869080-102869080 on Build GRCh38
rs545449702 Chr.4: 102869001-102869001 on Build GRCh38
rs776281350 Chr.4: 102869055-102869055 on Build GRCh38
rs1057132229 Chr.4: 102869052-102869052 on Build GRCh38
rs756961402 Chr.4: 102869126-102869126 on Build GRCh38
rs557515732 Chr.4: 102869083-102869083 on Build GRCh38
rs775480110 Chr.4: 102869080-102869080 on Build GRCh38
rs772041686 Chr.4: 102869048-102869049 on Build GRCh38
rs773631848 Chr.4: 102869152-102869152 on Build GRCh38
rs955276354 Chr.4: 102868987-102868987 on Build GRCh38
rs933324735 Chr.4: 102869005-102869005 on Build GRCh38
rs548270434 Chr.4: 102869026-102869026 on Build GRCh38
rs961514778 Chr.4: 102869101-102869101 on Build GRCh38
rs544395415 Chr.4: 102868967-102868967 on Build GRCh38
rs745626265 Chr.4: 102869046-102869048 on Build GRCh38
rs967263853 Chr.4: 102868958-102868958 on Build GRCh38
rs539010424 Chr.4: 102869074-102869074 on Build GRCh38
rs779629996 Chr.4: 102869143-102869143 on Build GRCh38
rs769308187 Chr.4: 102869057-102869057 on Build GRCh38
rs946494503 Chr.4: 102868998-102868998 on Build GRCh38

View Full Product Details