Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174812304-174812549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHN1
Entrez Gene ID
1123
Gene Name
chimerin 1
Gene Aliases
ARHGAP2, CHN, DURS2, NC, RHOGAP2
UniGene
Hs.380138
Gene Chromosome Location
Chr.2: 174799314-175005379 on Build GRCh38
Transcripts
NM_001025201.3 NM_001206602.1 NM_001822.5 NR_038133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628498 c.438G>A p.T146T Chr.2: 174812382-174812382 on Build GRCh38
4763626 c.409G>C p.V137L Chr.2: 174812411-174812411 on Build GRCh38
4883356 c.886G>T p.G296C Chr.2: 174812309-174812309 on Build GRCh38
1613719 c.781C>T p.H261Y Chr.2: 174812414-174812414 on Build GRCh38
1590899 c.441G>A p.T147T Chr.2: 174812379-174812379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139467204 Chr.2: 174812460-174812460 on Build GRCh38
rs778034298 Chr.2: 174812424-174812424 on Build GRCh38
rs57759612 Chr.2: 174812500-174812503 on Build GRCh38
rs121912797 Chr.2: 174812440-174812440 on Build GRCh38
rs756331019 Chr.2: 174812411-174812411 on Build GRCh38
rs891372818 Chr.2: 174812496-174812499 on Build GRCh38
rs776373199 Chr.2: 174812509-174812509 on Build GRCh38
rs771614355 Chr.2: 174812341-174812341 on Build GRCh38
rs751363901 Chr.2: 174812407-174812407 on Build GRCh38
rs199663916 Chr.2: 174812382-174812382 on Build GRCh38
rs769304555 Chr.2: 174812512-174812512 on Build GRCh38
rs528214646 Chr.2: 174812379-174812379 on Build GRCh38
rs757445182 Chr.2: 174812450-174812450 on Build GRCh38
rs760305779 Chr.2: 174812376-174812376 on Build GRCh38
rs749422915 Chr.2: 174812449-174812449 on Build GRCh38
rs387906600 Chr.2: 174812441-174812441 on Build GRCh38
rs772571308 Chr.2: 174812350-174812350 on Build GRCh38
rs768849610 Chr.2: 174812508-174812508 on Build GRCh38
rs35844789 Chr.2: 174812504-174812505 on Build GRCh38
rs746338135 Chr.2: 174812348-174812348 on Build GRCh38
rs766218478 Chr.2: 174812396-174812396 on Build GRCh38
rs761404391 Chr.2: 174812380-174812380 on Build GRCh38
rs775787683 Chr.2: 174812470-174812470 on Build GRCh38
rs772694741 Chr.2: 174812516-174812516 on Build GRCh38
rs561047964 Chr.2: 174812343-174812343 on Build GRCh38
rs746393214 Chr.2: 174812452-174812452 on Build GRCh38
rs761385486 Chr.2: 174812511-174812511 on Build GRCh38
rs747373088 Chr.2: 174812499-174812499 on Build GRCh38
rs749356855 Chr.2: 174812335-174812335 on Build GRCh38
rs373546784 Chr.2: 174812375-174812375 on Build GRCh38
rs764684387 Chr.2: 174812381-174812381 on Build GRCh38
rs564758271 Chr.2: 174812383-174812383 on Build GRCh38

View Full Product Details