Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33238014-33238249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA1
Entrez Gene ID
51297
Gene Name
BPI fold containing family A member 1
Gene Aliases
LUNX, NASG, PLUNC, SPLUNC1, SPURT, bA49G10.5
UniGene
Hs.211092
Gene Chromosome Location
Chr.20: 33235996-33243309 on Build GRCh38
Transcripts
NM_001243193.1 NM_016583.3 NM_130852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3840771 c.187G>A p.G63S Chr.20: 33238081-33238081 on Build GRCh38
3707710 c.186G>A p.G62G Chr.20: 33238080-33238080 on Build GRCh38
5464900 c.178C>T p.L60L Chr.20: 33238072-33238072 on Build GRCh38
6361482 c.184G>T p.G62W Chr.20: 33238078-33238078 on Build GRCh38
1411183 c.273G>T p.L91L Chr.20: 33238167-33238167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760068411 Chr.20: 33238177-33238177 on Build GRCh38
rs148950292 Chr.20: 33238182-33238182 on Build GRCh38
rs141888881 Chr.20: 33238203-33238203 on Build GRCh38
rs763520922 Chr.20: 33238186-33238186 on Build GRCh38
rs200411187 Chr.20: 33238158-33238158 on Build GRCh38
rs765939202 Chr.20: 33238181-33238181 on Build GRCh38
rs755458781 Chr.20: 33238087-33238087 on Build GRCh38
rs762616289 Chr.20: 33238059-33238059 on Build GRCh38
rs771763926 Chr.20: 33238171-33238171 on Build GRCh38
rs745983430 Chr.20: 33238127-33238127 on Build GRCh38
rs750695894 Chr.20: 33238210-33238210 on Build GRCh38
rs767007833 Chr.20: 33238074-33238074 on Build GRCh38
rs774194223 Chr.20: 33238160-33238160 on Build GRCh38
rs775322909 Chr.20: 33238054-33238054 on Build GRCh38
rs929164465 Chr.20: 33238104-33238104 on Build GRCh38
rs781522009 Chr.20: 33238206-33238206 on Build GRCh38
rs765705916 Chr.20: 33238092-33238092 on Build GRCh38
rs753282962 Chr.20: 33238097-33238097 on Build GRCh38
rs749261788 Chr.20: 33238142-33238142 on Build GRCh38
rs769667692 Chr.20: 33238052-33238052 on Build GRCh38
rs6059183 Chr.20: 33238221-33238221 on Build GRCh38
rs770401299 Chr.20: 33238163-33238163 on Build GRCh38
rs749989724 Chr.20: 33238086-33238086 on Build GRCh38
rs2752909 Chr.20: 33238053-33238053 on Build GRCh38
rs757621515 Chr.20: 33238111-33238111 on Build GRCh38
rs1054750883 Chr.20: 33238215-33238215 on Build GRCh38
rs776858314 Chr.20: 33238038-33238038 on Build GRCh38
rs752029139 Chr.20: 33238194-33238194 on Build GRCh38
rs964973272 Chr.20: 33238055-33238055 on Build GRCh38
rs375396851 Chr.20: 33238061-33238061 on Build GRCh38
rs140964610 Chr.20: 33238123-33238123 on Build GRCh38
rs143582801 Chr.20: 33238110-33238110 on Build GRCh38
rs758644147 Chr.20: 33238109-33238109 on Build GRCh38
rs145787306 Chr.20: 33238120-33238120 on Build GRCh38
rs773813670 Chr.20: 33238065-33238065 on Build GRCh38
rs368242286 Chr.20: 33238173-33238173 on Build GRCh38
rs759945968 Chr.20: 33238071-33238071 on Build GRCh38
rs764363602 Chr.20: 33238192-33238192 on Build GRCh38
rs779807290 Chr.20: 33238137-33238137 on Build GRCh38
rs768265114 Chr.20: 33238143-33238143 on Build GRCh38
rs769945839 Chr.20: 33238131-33238131 on Build GRCh38
rs745797507 Chr.20: 33238046-33238046 on Build GRCh38

View Full Product Details