Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36805359-36805590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPGS2
Entrez Gene ID
25941
Gene Name
tubulin polyglutamylase complex subunit 2
Gene Aliases
C18orf10, HMFN0601, L17, PGs2
UniGene
Hs.436636
Gene Chromosome Location
Chr.18: 36777647-36829393 on Build GRCh38
Transcripts
NM_001271949.1 NM_001271950.1 NM_001271951.1 NM_001271952.1 NM_001271953.1 NM_001271954.1 NM_001271955.1 NM_001271956.1 NM_015476.3 XM_005258242.3 XM_011525917.2 XM_017025699.1 XM_017025700.1 XM_017025701.1 XM_017025702.1 XM_017025703.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6195524 c.327G>A p.M109I Chr.18: 36805429-36805429 on Build GRCh38
1611225 c.382G>T p.A128S Chr.18: 36805374-36805374 on Build GRCh38
6148652 c.348C>T p.P116P Chr.18: 36805408-36805408 on Build GRCh38
6148653 c.348C>T p.P116P Chr.18: 36805408-36805408 on Build GRCh38
6195523 c.327G>A p.M109I Chr.18: 36805429-36805429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368837189 Chr.18: 36805517-36805517 on Build GRCh38
rs764319917 Chr.18: 36805477-36805477 on Build GRCh38
rs978891982 Chr.18: 36805423-36805423 on Build GRCh38
rs756601472 Chr.18: 36805501-36805501 on Build GRCh38
rs7234328 Chr.18: 36805386-36805386 on Build GRCh38
rs766350480 Chr.18: 36805537-36805537 on Build GRCh38
rs1039683760 Chr.18: 36805560-36805560 on Build GRCh38
rs372399401 Chr.18: 36805525-36805525 on Build GRCh38
rs771801742 Chr.18: 36805513-36805513 on Build GRCh38
rs768068824 Chr.18: 36805504-36805504 on Build GRCh38
rs752817812 Chr.18: 36805462-36805462 on Build GRCh38
rs773882784 Chr.18: 36805443-36805443 on Build GRCh38
rs780607810 Chr.18: 36805503-36805503 on Build GRCh38
rs117586096 Chr.18: 36805562-36805562 on Build GRCh38
rs963675605 Chr.18: 36805410-36805410 on Build GRCh38
rs779668155 Chr.18: 36805518-36805527 on Build GRCh38
rs114281017 Chr.18: 36805396-36805396 on Build GRCh38
rs760662956 Chr.18: 36805455-36805455 on Build GRCh38
rs761615059 Chr.18: 36805448-36805448 on Build GRCh38
rs746281969 Chr.18: 36805496-36805496 on Build GRCh38
rs746406454 Chr.18: 36805414-36805414 on Build GRCh38
rs774009358 Chr.18: 36805507-36805507 on Build GRCh38
rs943638831 Chr.18: 36805546-36805546 on Build GRCh38
rs757602186 Chr.18: 36805485-36805485 on Build GRCh38
rs773178578 Chr.18: 36805452-36805452 on Build GRCh38
rs374431674 Chr.18: 36805418-36805418 on Build GRCh38
rs776721183 Chr.18: 36805552-36805552 on Build GRCh38
rs149427975 Chr.18: 36805431-36805431 on Build GRCh38
rs781687680 Chr.18: 36805398-36805398 on Build GRCh38
rs562703180 Chr.18: 36805392-36805392 on Build GRCh38
rs748855421 Chr.18: 36805423-36805423 on Build GRCh38
rs767242428 Chr.18: 36805449-36805449 on Build GRCh38
rs758677668 Chr.18: 36805387-36805387 on Build GRCh38
rs777922899 Chr.18: 36805389-36805389 on Build GRCh38
rs773125577 Chr.18: 36805516-36805516 on Build GRCh38
rs747720769 Chr.18: 36805512-36805512 on Build GRCh38
rs751751150 Chr.18: 36805479-36805479 on Build GRCh38
rs766480030 Chr.18: 36805459-36805459 on Build GRCh38
rs144816782 Chr.18: 36805489-36805489 on Build GRCh38
rs758411280 Chr.18: 36805473-36805473 on Build GRCh38
rs774960292 Chr.18: 36805419-36805419 on Build GRCh38
rs1029302516 Chr.18: 36805467-36805467 on Build GRCh38

View Full Product Details