Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57474525-57474792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP9
Entrez Gene ID
64333
Gene Name
Rho GTPase activating protein 9
Gene Aliases
10C, RGL1
UniGene
Hs.437126
Gene Chromosome Location
Chr.12: 57472255-57488814 on Build GRCh38
Transcripts
NM_001080156.2 NM_001080157.1 NM_001319850.1 NM_001319851.1 NM_001319852.1 NM_032496.3 XM_005269083.2 XM_005269084.2 XM_005269085.2 XM_011538656.2 XM_011538657.2 XM_011538658.2 XM_011538659.2 XM_017019800.1 XM_017019801.1 XM_017019802.1 XM_017019803.1 XM_017019804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6302687 c.1779G>T p.V593V Chr.12: 57474633-57474633 on Build GRCh38
3704300 c.1771T>G p.F591V Chr.12: 57474641-57474641 on Build GRCh38
6346383 c.1757T>A p.V586D Chr.12: 57474655-57474655 on Build GRCh38
7107043 c.1724G>A p.G575D Chr.12: 57474688-57474688 on Build GRCh38
5782693 c.1769G>A p.R590H Chr.12: 57474643-57474643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541605179 Chr.12: 57474577-57474577 on Build GRCh38
rs962667515 Chr.12: 57474556-57474556 on Build GRCh38
rs766890985 Chr.12: 57474575-57474575 on Build GRCh38
rs753744407 Chr.12: 57474598-57474598 on Build GRCh38
rs2292619 Chr.12: 57474690-57474690 on Build GRCh38
rs575341846 Chr.12: 57474725-57474725 on Build GRCh38
rs373002969 Chr.12: 57474616-57474616 on Build GRCh38
rs545453716 Chr.12: 57474755-57474755 on Build GRCh38
rs370706224 Chr.12: 57474678-57474678 on Build GRCh38
rs553244231 Chr.12: 57474579-57474579 on Build GRCh38
rs1016864343 Chr.12: 57474551-57474551 on Build GRCh38
rs757095950 Chr.12: 57474601-57474601 on Build GRCh38
rs778948150 Chr.12: 57474606-57474606 on Build GRCh38
rs901752267 Chr.12: 57474759-57474759 on Build GRCh38
rs750385634 Chr.12: 57474723-57474723 on Build GRCh38
rs917448795 Chr.12: 57474734-57474734 on Build GRCh38
rs1044489975 Chr.12: 57474730-57474730 on Build GRCh38
rs750286030 Chr.12: 57474607-57474607 on Build GRCh38
rs989037908 Chr.12: 57474546-57474546 on Build GRCh38
rs768928515 Chr.12: 57474647-57474647 on Build GRCh38
rs771617741 Chr.12: 57474675-57474675 on Build GRCh38
rs781373940 Chr.12: 57474656-57474656 on Build GRCh38
rs561435439 Chr.12: 57474672-57474672 on Build GRCh38
rs371682585 Chr.12: 57474566-57474566 on Build GRCh38
rs760338083 Chr.12: 57474587-57474587 on Build GRCh38
rs770018068 Chr.12: 57474669-57474669 on Build GRCh38
rs575082970 Chr.12: 57474618-57474618 on Build GRCh38
rs761698560 Chr.12: 57474712-57474712 on Build GRCh38
rs776390646 Chr.12: 57474695-57474695 on Build GRCh38
rs375113199 Chr.12: 57474567-57474567 on Build GRCh38
rs376622753 Chr.12: 57474644-57474644 on Build GRCh38
rs747233606 Chr.12: 57474633-57474633 on Build GRCh38
rs763835138 Chr.12: 57474590-57474590 on Build GRCh38
rs760248150 Chr.12: 57474679-57474679 on Build GRCh38
rs1048775810 Chr.12: 57474683-57474683 on Build GRCh38
rs748556613 Chr.12: 57474661-57474661 on Build GRCh38
rs773793665 Chr.12: 57474671-57474671 on Build GRCh38
rs557077108 Chr.12: 57474715-57474715 on Build GRCh38
rs61935685 Chr.12: 57474610-57474610 on Build GRCh38

View Full Product Details