Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151187291-151187560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC69
Entrez Gene ID
26112
Gene Name
coiled-coil domain containing 69
Gene Aliases
-
UniGene
Hs.655336
Gene Chromosome Location
Chr.5: 151181052-151224093 on Build GRCh38
Transcripts
NM_015621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3852862 c.385A>G p.T129A Chr.5: 151187394-151187394 on Build GRCh38
1672048 c.383C>A p.S128Y Chr.5: 151187396-151187396 on Build GRCh38
4433588 c.390G>T p.Q130H Chr.5: 151187389-151187389 on Build GRCh38
6622384 c.365C>A p.S122Y Chr.5: 151187414-151187414 on Build GRCh38
449129 c.393+1G>T p.? Chr.5: 151187385-151187385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752376662 Chr.5: 151187505-151187505 on Build GRCh38
rs147336268 Chr.5: 151187424-151187424 on Build GRCh38
rs757094623 Chr.5: 151187352-151187354 on Build GRCh38
rs762794491 Chr.5: 151187472-151187472 on Build GRCh38
rs36036156 Chr.5: 151187513-151187514 on Build GRCh38
rs751437294 Chr.5: 151187427-151187427 on Build GRCh38
rs777392184 Chr.5: 151187444-151187444 on Build GRCh38
rs933644296 Chr.5: 151187363-151187363 on Build GRCh38
rs746175735 Chr.5: 151187453-151187453 on Build GRCh38
rs1037571768 Chr.5: 151187450-151187450 on Build GRCh38
rs761728854 Chr.5: 151187349-151187349 on Build GRCh38
rs150829162 Chr.5: 151187376-151187376 on Build GRCh38
rs115540962 Chr.5: 151187408-151187408 on Build GRCh38
rs116701374 Chr.5: 151187454-151187454 on Build GRCh38
rs538272458 Chr.5: 151187522-151187522 on Build GRCh38
rs376335487 Chr.5: 151187368-151187368 on Build GRCh38
rs796757702 Chr.5: 151187490-151187490 on Build GRCh38
rs386693598 Chr.5: 151187454-151187456 on Build GRCh38
rs35221501 Chr.5: 151187489-151187489 on Build GRCh38
rs758085908 Chr.5: 151187361-151187361 on Build GRCh38
rs78512767 Chr.5: 151187325-151187325 on Build GRCh38
rs754663499 Chr.5: 151187434-151187434 on Build GRCh38
rs774442502 Chr.5: 151187500-151187500 on Build GRCh38
rs61745454 Chr.5: 151187456-151187456 on Build GRCh38
rs200423694 Chr.5: 151187466-151187466 on Build GRCh38
rs745842660 Chr.5: 151187364-151187365 on Build GRCh38
rs367923607 Chr.5: 151187340-151187340 on Build GRCh38
rs539734088 Chr.5: 151187463-151187463 on Build GRCh38
rs138290590 Chr.5: 151187440-151187440 on Build GRCh38
rs750377297 Chr.5: 151187355-151187355 on Build GRCh38
rs866016314 Chr.5: 151187356-151187356 on Build GRCh38
rs778103483 Chr.5: 151187385-151187385 on Build GRCh38
rs887429293 Chr.5: 151187459-151187459 on Build GRCh38
rs752844296 Chr.5: 151187345-151187345 on Build GRCh38
rs369726489 Chr.5: 151187375-151187375 on Build GRCh38
rs779135902 Chr.5: 151187452-151187452 on Build GRCh38
rs749825933 Chr.5: 151187391-151187391 on Build GRCh38
rs760684238 Chr.5: 151187421-151187421 on Build GRCh38
rs201014200 Chr.5: 151187350-151187350 on Build GRCh38
rs772342301 Chr.5: 151187409-151187409 on Build GRCh38
rs371053110 Chr.5: 151187482-151187482 on Build GRCh38
rs779917411 Chr.5: 151187367-151187367 on Build GRCh38
rs780932993 Chr.5: 151187439-151187439 on Build GRCh38
rs774819545 Chr.5: 151187406-151187406 on Build GRCh38
rs397702931 Chr.5: 151187514-151187515 on Build GRCh38
rs780948098 Chr.5: 151187360-151187361 on Build GRCh38
rs775647410 Chr.5: 151187455-151187455 on Build GRCh38
rs780843491 Chr.5: 151187369-151187369 on Build GRCh38
rs200558334 Chr.5: 151187504-151187504 on Build GRCh38
rs61745455 Chr.5: 151187449-151187449 on Build GRCh38
rs142006070 Chr.5: 151187411-151187411 on Build GRCh38
rs549592183 Chr.5: 151187426-151187426 on Build GRCh38
rs755871464 Chr.5: 151187506-151187506 on Build GRCh38
rs771177955 Chr.5: 151187396-151187396 on Build GRCh38
rs755637802 Chr.5: 151187442-151187442 on Build GRCh38

View Full Product Details