Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82831861-82832118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBCD
Entrez Gene ID
6904
Gene Name
tubulin folding cofactor D
Gene Aliases
SSD-1, tfcD
UniGene
Hs.464391
Gene Chromosome Location
Chr.17: 82752048-82943186 on Build GRCh38
Transcripts
NM_005993.4 XM_005256396.4 XM_006722290.2 XM_011523589.1 XM_011523590.1 XM_011523591.1 XM_011523593.1 XM_017024987.1 XM_017024988.1 XM_017024990.1 NM_024702.2

Gene Symbol
ZNF750
Entrez Gene ID
79755
Gene Name
zinc finger protein 750
Gene Aliases
ZFP750
UniGene
Hs.464391
Gene Chromosome Location
Chr.17: 82829434-82840055 on Build GRCh38
Transcripts
NM_005993.4 XM_005256396.4 XM_006722290.2 XM_011523589.1 XM_011523590.1 XM_011523591.1 XM_011523593.1 XM_017024987.1 XM_017024988.1 XM_017024990.1 NM_024702.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5111542 c.573C>A p.A191A Chr.17: 82831882-82831882 on Build GRCh38
6995138 c.400delA p.R134fs*232 Chr.17: 82832055-82832055 on Build GRCh38
4070957 c.429G>A p.P143P Chr.17: 82832026-82832026 on Build GRCh38
1271760 c.538G>A p.E180K Chr.17: 82831917-82831917 on Build GRCh38
5628261 c.449A>C p.Q150P Chr.17: 82832006-82832006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149980253 Chr.17: 82831982-82831982 on Build GRCh38
rs916058616 Chr.17: 82832049-82832049 on Build GRCh38
rs147701658 Chr.17: 82832014-82832014 on Build GRCh38
rs759008982 Chr.17: 82831959-82831959 on Build GRCh38
rs776475239 Chr.17: 82832097-82832097 on Build GRCh38
rs773981424 Chr.17: 82831996-82831996 on Build GRCh38
rs539319902 Chr.17: 82832077-82832077 on Build GRCh38
rs146491968 Chr.17: 82832031-82832031 on Build GRCh38
rs936983261 Chr.17: 82831991-82831991 on Build GRCh38
rs965277222 Chr.17: 82831922-82831922 on Build GRCh38
rs371018840 Chr.17: 82832032-82832032 on Build GRCh38
rs753964912 Chr.17: 82831882-82831882 on Build GRCh38
rs769621145 Chr.17: 82831926-82831926 on Build GRCh38
rs142698629 Chr.17: 82831950-82831950 on Build GRCh38
rs768666262 Chr.17: 82831928-82831928 on Build GRCh38
rs200912015 Chr.17: 82831921-82831921 on Build GRCh38
rs948959982 Chr.17: 82831918-82831918 on Build GRCh38
rs766625742 Chr.17: 82831940-82831940 on Build GRCh38
rs142460137 Chr.17: 82832026-82832026 on Build GRCh38
rs374227498 Chr.17: 82831956-82831956 on Build GRCh38
rs898850065 Chr.17: 82832084-82832084 on Build GRCh38
rs755072404 Chr.17: 82831951-82831951 on Build GRCh38
rs752878414 Chr.17: 82832027-82832027 on Build GRCh38
rs766535915 Chr.17: 82832022-82832022 on Build GRCh38
rs753147570 Chr.17: 82831957-82831957 on Build GRCh38
rs374920189 Chr.17: 82832085-82832085 on Build GRCh38
rs200058922 Chr.17: 82831933-82831933 on Build GRCh38
rs748948272 Chr.17: 82832054-82832054 on Build GRCh38
rs772336681 Chr.17: 82831915-82831915 on Build GRCh38
rs968728616 Chr.17: 82831998-82831998 on Build GRCh38
rs746747831 Chr.17: 82832094-82832094 on Build GRCh38
rs552853276 Chr.17: 82831904-82831904 on Build GRCh38
rs977516372 Chr.17: 82832010-82832010 on Build GRCh38
rs748495076 Chr.17: 82831913-82831913 on Build GRCh38
rs1056704898 Chr.17: 82832011-82832011 on Build GRCh38
rs755442581 Chr.17: 82831890-82831890 on Build GRCh38
rs35867846 Chr.17: 82831960-82831960 on Build GRCh38
rs773151722 Chr.17: 82832092-82832092 on Build GRCh38
rs761646138 Chr.17: 82832000-82832000 on Build GRCh38
rs754719607 Chr.17: 82832063-82832063 on Build GRCh38
rs187135973 Chr.17: 82831881-82831881 on Build GRCh38
rs371042619 Chr.17: 82832096-82832096 on Build GRCh38
rs35182876 Chr.17: 82832090-82832090 on Build GRCh38
rs750293670 Chr.17: 82831939-82831939 on Build GRCh38
rs778267081 Chr.17: 82832069-82832069 on Build GRCh38
rs894728784 Chr.17: 82831894-82831894 on Build GRCh38
rs760490074 Chr.17: 82832019-82832019 on Build GRCh38
rs535551971 Chr.17: 82832009-82832009 on Build GRCh38
rs779876008 Chr.17: 82831981-82831981 on Build GRCh38
rs1022071365 Chr.17: 82831995-82831995 on Build GRCh38
rs1037944628 Chr.17: 82832079-82832079 on Build GRCh38
rs1057351778 Chr.17: 82831884-82831884 on Build GRCh38
rs953376583 Chr.17: 82831916-82831916 on Build GRCh38
rs768060518 Chr.17: 82831983-82831983 on Build GRCh38
rs754071746 Chr.17: 82832023-82832023 on Build GRCh38
rs757711155 Chr.17: 82832051-82832051 on Build GRCh38
rs147504960 Chr.17: 82831930-82831930 on Build GRCh38
rs751236472 Chr.17: 82831927-82831927 on Build GRCh38
rs368920040 Chr.17: 82831962-82831962 on Build GRCh38
rs774173103 Chr.17: 82831929-82831929 on Build GRCh38
rs369588218 Chr.17: 82832042-82832042 on Build GRCh38
rs374434732 Chr.17: 82831917-82831917 on Build GRCh38
rs751924669 Chr.17: 82832038-82832038 on Build GRCh38

View Full Product Details