Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 34862843-34863096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC23L
Entrez Gene ID
153657
Gene Name
tetratricopeptide repeat domain 23 like
Gene Aliases
MC25-1
UniGene
Hs.435742
Gene Chromosome Location
Chr.5: 34839164-34900511 on Build GRCh38
Transcripts
NM_001317949.1 NM_144725.3 XM_011513984.2 XM_011513985.2 XM_011513987.2 XM_017009119.1 XM_017009120.1 XM_017009121.1 XM_017009122.1 XM_017009123.1 XM_017009124.1 XM_017009125.1 XM_017009126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5939480 c.385C>T p.P129S Chr.5: 34862903-34862903 on Build GRCh38
3761209 c.411A>T p.T137T Chr.5: 34862929-34862929 on Build GRCh38
5935069 c.536G>A p.R179H Chr.5: 34863054-34863054 on Build GRCh38
7424599 c.454T>G p.S152A Chr.5: 34862972-34862972 on Build GRCh38
242668 c.514G>A p.V172M Chr.5: 34863032-34863032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373203201 Chr.5: 34862887-34862887 on Build GRCh38
rs746574162 Chr.5: 34863006-34863006 on Build GRCh38
rs369127609 Chr.5: 34862876-34862876 on Build GRCh38
rs964056508 Chr.5: 34862882-34862882 on Build GRCh38
rs767386776 Chr.5: 34863055-34863055 on Build GRCh38
rs754675463 Chr.5: 34862933-34862933 on Build GRCh38
rs754439182 Chr.5: 34863055-34863056 on Build GRCh38
rs753393310 Chr.5: 34863077-34863077 on Build GRCh38
rs775944579 Chr.5: 34862877-34862877 on Build GRCh38
rs3906383 Chr.5: 34862929-34862929 on Build GRCh38
rs748696306 Chr.5: 34862987-34862987 on Build GRCh38
rs755723416 Chr.5: 34862968-34862968 on Build GRCh38
rs375509802 Chr.5: 34863074-34863074 on Build GRCh38
rs769004166 Chr.5: 34863016-34863016 on Build GRCh38
rs762876670 Chr.5: 34863049-34863049 on Build GRCh38
rs377261919 Chr.5: 34862891-34862891 on Build GRCh38
rs751435638 Chr.5: 34863053-34863053 on Build GRCh38
rs117051837 Chr.5: 34862906-34862906 on Build GRCh38
rs1002719618 Chr.5: 34862881-34862881 on Build GRCh38
rs752443931 Chr.5: 34863057-34863057 on Build GRCh38
rs765395457 Chr.5: 34863042-34863042 on Build GRCh38
rs761947724 Chr.5: 34862899-34862899 on Build GRCh38
rs766356998 Chr.5: 34863050-34863050 on Build GRCh38
rs750215899 Chr.5: 34862881-34862881 on Build GRCh38
rs746342208 Chr.5: 34862869-34862869 on Build GRCh38
rs368660770 Chr.5: 34863047-34863047 on Build GRCh38
rs747412266 Chr.5: 34862998-34862998 on Build GRCh38
rs776878152 Chr.5: 34863031-34863031 on Build GRCh38
rs781045694 Chr.5: 34862936-34862936 on Build GRCh38
rs374310226 Chr.5: 34863032-34863032 on Build GRCh38
rs35769440 Chr.5: 34862975-34862975 on Build GRCh38
rs368031020 Chr.5: 34862976-34862976 on Build GRCh38
rs779652913 Chr.5: 34862986-34862987 on Build GRCh38
rs34566695 Chr.5: 34862967-34862967 on Build GRCh38
rs755852616 Chr.5: 34863072-34863072 on Build GRCh38
rs185627068 Chr.5: 34862922-34862922 on Build GRCh38
rs897985800 Chr.5: 34863001-34863001 on Build GRCh38
rs899125002 Chr.5: 34862994-34862994 on Build GRCh38
rs775793618 Chr.5: 34862995-34862995 on Build GRCh38
rs571614688 Chr.5: 34862989-34862989 on Build GRCh38
rs758057412 Chr.5: 34862974-34862977 on Build GRCh38
rs371992268 Chr.5: 34863054-34863054 on Build GRCh38
rs369799222 Chr.5: 34863076-34863076 on Build GRCh38

View Full Product Details