Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 27951942-27952199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH2
Entrez Gene ID
1000
Gene Name
cadherin 2
Gene Aliases
CD325, CDHN, CDw325, NCAD
UniGene
Hs.464829
Gene Chromosome Location
Chr.18: 27932878-28177446 on Build GRCh38
Transcripts
NM_001308176.1 NM_001792.4 XM_011525788.1 XM_017025514.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
378241 c.2710G>T p.G904C Chr.18: 27952164-27952164 on Build GRCh38
385442 c.2678G>A p.R893Q Chr.18: 27952196-27952196 on Build GRCh38
4071663 c.2690T>A p.L897H Chr.18: 27952184-27952184 on Build GRCh38
4130944 c.2712T>G p.G904G Chr.18: 27952162-27952162 on Build GRCh38
987321 c.2714A>G p.D905G Chr.18: 27952160-27952160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747297620 Chr.18: 27952118-27952118 on Build GRCh38
rs765935402 Chr.18: 27952111-27952115 on Build GRCh38
rs199985888 Chr.18: 27952008-27952008 on Build GRCh38
rs778622999 Chr.18: 27952135-27952136 on Build GRCh38
rs994106390 Chr.18: 27951988-27951988 on Build GRCh38
rs759051449 Chr.18: 27952177-27952177 on Build GRCh38
rs761462195 Chr.18: 27952142-27952142 on Build GRCh38
rs201220952 Chr.18: 27951996-27951996 on Build GRCh38
rs753397750 Chr.18: 27952120-27952123 on Build GRCh38
rs200446668 Chr.18: 27952020-27952020 on Build GRCh38
rs540265163 Chr.18: 27952000-27952000 on Build GRCh38
rs779405421 Chr.18: 27952102-27952102 on Build GRCh38
rs768651826 Chr.18: 27952133-27952133 on Build GRCh38
rs771948589 Chr.18: 27952145-27952145 on Build GRCh38
rs202235245 Chr.18: 27952016-27952016 on Build GRCh38
rs758813164 Chr.18: 27952114-27952114 on Build GRCh38
rs891173638 Chr.18: 27952098-27952098 on Build GRCh38
rs200246655 Chr.18: 27952163-27952163 on Build GRCh38
rs754516111 Chr.18: 27952131-27952132 on Build GRCh38
rs201787083 Chr.18: 27952121-27952121 on Build GRCh38
rs201833801 Chr.18: 27952002-27952002 on Build GRCh38
rs1039649370 Chr.18: 27952139-27952139 on Build GRCh38
rs760441394 Chr.18: 27952149-27952149 on Build GRCh38
rs765791790 Chr.18: 27952161-27952161 on Build GRCh38
rs752993150 Chr.18: 27952109-27952109 on Build GRCh38
rs776060409 Chr.18: 27952176-27952176 on Build GRCh38
rs774465203 Chr.18: 27952135-27952135 on Build GRCh38
rs989076290 Chr.18: 27952148-27952148 on Build GRCh38
rs781482987 Chr.18: 27952127-27952127 on Build GRCh38
rs11564422 Chr.18: 27952116-27952116 on Build GRCh38
rs772705778 Chr.18: 27952147-27952147 on Build GRCh38
rs745961532 Chr.18: 27952128-27952128 on Build GRCh38
rs774735339 Chr.18: 27952055-27952055 on Build GRCh38
rs200920347 Chr.18: 27952013-27952013 on Build GRCh38

View Full Product Details