Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139745665-139745932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11C
Entrez Gene ID
286410
Gene Name
ATPase phospholipid transporting 11C
Gene Aliases
ATPIG, ATPIQ
UniGene
Hs.88252
Gene Chromosome Location
Chr.X: 139726346-139933083 on Build GRCh38
Transcripts
NM_001010986.2 NM_173694.4 XM_017029439.1 XM_017029440.1 XM_017029441.1 XM_017029442.1 XM_017029443.1 XM_017029444.1 XM_017029445.1 XM_017029446.1 XM_017029447.1 XM_017029448.1 XM_017029449.1

SNP Detail

SNP ID Chromosome Location
rs746593846 Chr.X: 139745814-139745814 on Build GRCh38
rs749194856 Chr.X: 139745863-139745863 on Build GRCh38
rs752065947 Chr.X: 139745705-139745705 on Build GRCh38
rs756953682 Chr.X: 139745823-139745823 on Build GRCh38
rs994648613 Chr.X: 139745873-139745873 on Build GRCh38
rs747028728 Chr.X: 139745876-139745876 on Build GRCh38
rs772422546 Chr.X: 139745719-139745719 on Build GRCh38
rs761382190 Chr.X: 139745875-139745876 on Build GRCh38
rs773425394 Chr.X: 139745698-139745698 on Build GRCh38
rs944483041 Chr.X: 139745763-139745763 on Build GRCh38
rs987558525 Chr.X: 139745745-139745745 on Build GRCh38
rs1048415175 Chr.X: 139745892-139745892 on Build GRCh38
rs765746541 Chr.X: 139745749-139745749 on Build GRCh38
rs12849115 Chr.X: 139745780-139745780 on Build GRCh38
rs745623704 Chr.X: 139745838-139745838 on Build GRCh38
rs12843381 Chr.X: 139745720-139745720 on Build GRCh38
rs55724992 Chr.X: 139745781-139745781 on Build GRCh38
rs755385365 Chr.X: 139745729-139745729 on Build GRCh38
rs1038307696 Chr.X: 139745911-139745911 on Build GRCh38
rs373196740 Chr.X: 139745760-139745760 on Build GRCh38
rs768417085 Chr.X: 139745782-139745782 on Build GRCh38
rs766656474 Chr.X: 139745700-139745700 on Build GRCh38
rs753180192 Chr.X: 139745754-139745754 on Build GRCh38
rs775449508 Chr.X: 139745854-139745854 on Build GRCh38
rs773162843 Chr.X: 139745901-139745901 on Build GRCh38
rs765588281 Chr.X: 139745876-139745876 on Build GRCh38
rs781037262 Chr.X: 139745831-139745831 on Build GRCh38
rs79484917 Chr.X: 139745856-139745856 on Build GRCh38
rs777164604 Chr.X: 139745778-139745778 on Build GRCh38

View Full Product Details