Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7054636-7054891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TADA2B
Entrez Gene ID
93624
Gene Name
transcriptional adaptor 2B
Gene Aliases
ADA2(beta), ADA2B
UniGene
Hs.518614
Gene Chromosome Location
Chr.4: 7043429-7057952 on Build GRCh38
Transcripts
NM_152293.2 XM_011513595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4830803 c.1055C>G p.S352* Chr.4: 7054846-7054846 on Build GRCh38
7142492 c.1017C>A p.F339L Chr.4: 7054808-7054808 on Build GRCh38
4125710 c.1072C>A p.L358I Chr.4: 7054863-7054863 on Build GRCh38
7144149 c.852C>G p.L284L Chr.4: 7054643-7054643 on Build GRCh38
6796150 c.932C>T p.A311V Chr.4: 7054723-7054723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755058859 Chr.4: 7054853-7054853 on Build GRCh38
rs772692077 Chr.4: 7054673-7054673 on Build GRCh38
rs372684985 Chr.4: 7054790-7054790 on Build GRCh38
rs746850955 Chr.4: 7054669-7054669 on Build GRCh38
rs773540665 Chr.4: 7054742-7054744 on Build GRCh38
rs750943990 Chr.4: 7054736-7054736 on Build GRCh38
rs199685832 Chr.4: 7054751-7054751 on Build GRCh38
rs369931391 Chr.4: 7054814-7054814 on Build GRCh38
rs768705454 Chr.4: 7054811-7054811 on Build GRCh38
rs761955767 Chr.4: 7054815-7054815 on Build GRCh38
rs775464029 Chr.4: 7054802-7054802 on Build GRCh38
rs377183651 Chr.4: 7054808-7054808 on Build GRCh38
rs767937296 Chr.4: 7054816-7054816 on Build GRCh38
rs371816393 Chr.4: 7054693-7054693 on Build GRCh38
rs767862945 Chr.4: 7054734-7054734 on Build GRCh38
rs557910360 Chr.4: 7054753-7054753 on Build GRCh38
rs773398504 Chr.4: 7054827-7054827 on Build GRCh38
rs777537320 Chr.4: 7054656-7054656 on Build GRCh38
rs777627029 Chr.4: 7054766-7054766 on Build GRCh38
rs757960745 Chr.4: 7054761-7054761 on Build GRCh38
rs376472841 Chr.4: 7054767-7054767 on Build GRCh38
rs774530772 Chr.4: 7054724-7054724 on Build GRCh38
rs539508764 Chr.4: 7054682-7054682 on Build GRCh38
rs764337259 Chr.4: 7054723-7054723 on Build GRCh38
rs963844224 Chr.4: 7054795-7054795 on Build GRCh38
rs762223816 Chr.4: 7054726-7054726 on Build GRCh38
rs761071358 Chr.4: 7054831-7054831 on Build GRCh38
rs780794883 Chr.4: 7054775-7054775 on Build GRCh38
rs752190983 Chr.4: 7054754-7054754 on Build GRCh38
rs775380828 Chr.4: 7054709-7054709 on Build GRCh38
rs914496603 Chr.4: 7054836-7054836 on Build GRCh38
rs368347599 Chr.4: 7054776-7054776 on Build GRCh38
rs766846018 Chr.4: 7054839-7054839 on Build GRCh38
rs745728932 Chr.4: 7054685-7054685 on Build GRCh38

View Full Product Details