Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142313069-142313326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRN1
Entrez Gene ID
54464
Gene Name
5'-3' exoribonuclease 1
Gene Aliases
SEP1
UniGene
Hs.435103
Gene Chromosome Location
Chr.3: 142306607-142448062 on Build GRCh38
Transcripts
NM_001282857.1 NM_019001.4 XM_011512919.2 XM_011512920.2 XM_011512922.2 XM_017006640.1 XM_017006641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3726243 c.4623G>T p.W1541C Chr.3: 142313170-142313170 on Build GRCh38
1617005 c.4631A>G p.Y1544C Chr.3: 142313162-142313162 on Build GRCh38
6440405 c.4654G>C p.A1552P Chr.3: 142313139-142313139 on Build GRCh38
5595574 c.4627C>T p.H1543Y Chr.3: 142313166-142313166 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879131062 Chr.3: 142313289-142313292 on Build GRCh38
rs369070014 Chr.3: 142313179-142313179 on Build GRCh38
rs546865136 Chr.3: 142313237-142313237 on Build GRCh38
rs200334821 Chr.3: 142313111-142313111 on Build GRCh38
rs776617685 Chr.3: 142313145-142313145 on Build GRCh38
rs746908761 Chr.3: 142313130-142313130 on Build GRCh38
rs769616091 Chr.3: 142313170-142313170 on Build GRCh38
rs369919016 Chr.3: 142313186-142313186 on Build GRCh38
rs932592266 Chr.3: 142313232-142313232 on Build GRCh38
rs768120876 Chr.3: 142313223-142313223 on Build GRCh38
rs113276810 Chr.3: 142313137-142313137 on Build GRCh38
rs969433208 Chr.3: 142313098-142313098 on Build GRCh38
rs941216799 Chr.3: 142313271-142313271 on Build GRCh38
rs984758476 Chr.3: 142313185-142313185 on Build GRCh38
rs773381241 Chr.3: 142313175-142313175 on Build GRCh38
rs777345660 Chr.3: 142313174-142313193 on Build GRCh38
rs770919461 Chr.3: 142313193-142313194 on Build GRCh38
rs748676042 Chr.3: 142313178-142313179 on Build GRCh38
rs377369694 Chr.3: 142313158-142313158 on Build GRCh38
rs769278192 Chr.3: 142313268-142313268 on Build GRCh38
rs768586814 Chr.3: 142313138-142313138 on Build GRCh38
rs747916274 Chr.3: 142313150-142313150 on Build GRCh38
rs934332267 Chr.3: 142313173-142313173 on Build GRCh38
rs776953029 Chr.3: 142313286-142313287 on Build GRCh38
rs770982211 Chr.3: 142313180-142313180 on Build GRCh38
rs779455342 Chr.3: 142313117-142313117 on Build GRCh38
rs759724311 Chr.3: 142313214-142313214 on Build GRCh38

View Full Product Details