Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60115388-60115621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCNX4
Entrez Gene ID
64430
Gene Name
pecanex homolog 4 (Drosophila)
Gene Aliases
C14orf135, FBP2, PCNXL4
UniGene
Hs.509499
Gene Chromosome Location
Chr.14: 60091276-60142976 on Build GRCh38
Transcripts
NM_022495.5 XM_005268009.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
698215 c.591A>G p.V197V Chr.14: 60115397-60115397 on Build GRCh38
267936 c.1320G>T p.M440I Chr.14: 60115424-60115424 on Build GRCh38
6282478 c.625G>A p.G209S Chr.14: 60115431-60115431 on Build GRCh38
2246664 c.1339C>T p.R447W Chr.14: 60115443-60115443 on Build GRCh38
267935 c.618G>T p.M206I Chr.14: 60115424-60115424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973297568 Chr.14: 60115504-60115504 on Build GRCh38
rs769252155 Chr.14: 60115501-60115501 on Build GRCh38
rs1040096777 Chr.14: 60115573-60115573 on Build GRCh38
rs184316913 Chr.14: 60115484-60115484 on Build GRCh38
rs764543492 Chr.14: 60115430-60115430 on Build GRCh38
rs535098016 Chr.14: 60115444-60115444 on Build GRCh38
rs773002851 Chr.14: 60115419-60115419 on Build GRCh38
rs929086401 Chr.14: 60115513-60115513 on Build GRCh38
rs565987164 Chr.14: 60115422-60115422 on Build GRCh38
rs749111462 Chr.14: 60115479-60115479 on Build GRCh38
rs901839979 Chr.14: 60115531-60115531 on Build GRCh38
rs777406702 Chr.14: 60115503-60115503 on Build GRCh38
rs889202630 Chr.14: 60115554-60115554 on Build GRCh38
rs748993311 Chr.14: 60115509-60115509 on Build GRCh38
rs754752275 Chr.14: 60115464-60115464 on Build GRCh38
rs781005229 Chr.14: 60115478-60115478 on Build GRCh38
rs761281541 Chr.14: 60115428-60115428 on Build GRCh38
rs944039327 Chr.14: 60115560-60115560 on Build GRCh38
rs748049557 Chr.14: 60115499-60115499 on Build GRCh38
rs754325608 Chr.14: 60115433-60115433 on Build GRCh38
rs562315792 Chr.14: 60115443-60115443 on Build GRCh38
rs1051690409 Chr.14: 60115539-60115539 on Build GRCh38
rs899947392 Chr.14: 60115586-60115586 on Build GRCh38
rs770431436 Chr.14: 60115537-60115537 on Build GRCh38
rs1016114337 Chr.14: 60115562-60115562 on Build GRCh38
rs775970110 Chr.14: 60115423-60115423 on Build GRCh38
rs751326164 Chr.14: 60115462-60115462 on Build GRCh38
rs1029033240 Chr.14: 60115543-60115543 on Build GRCh38
rs115609434 Chr.14: 60115558-60115558 on Build GRCh38
rs761813889 Chr.14: 60115434-60115434 on Build GRCh38
rs568638092 Chr.14: 60115524-60115524 on Build GRCh38
rs758607688 Chr.14: 60115451-60115451 on Build GRCh38

View Full Product Details