Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169740054-169740283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCTE3
Entrez Gene ID
6991
Gene Name
t-complex-associated-testis-expressed 3
Gene Aliases
TCTEX1D3
UniGene
Hs.733746
Gene Chromosome Location
Chr.6: 169725488-169751581 on Build GRCh38
Transcripts
NM_174910.1 XM_006715554.3 XM_011536093.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5900072 c.508G>A p.D170N Chr.6: 169740274-169740274 on Build GRCh38
6798917 c.558C>T p.Y186Y Chr.6: 169740224-169740224 on Build GRCh38
6847439 c.502A>T p.I168F Chr.6: 169740280-169740280 on Build GRCh38
4732844 c.588T>C p.Y196Y Chr.6: 169740194-169740194 on Build GRCh38
3381527 c.567G>A p.L189L Chr.6: 169740215-169740215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112863889 Chr.6: 169740236-169740236 on Build GRCh38
rs9383520 Chr.6: 169740088-169740088 on Build GRCh38
rs765765046 Chr.6: 169740250-169740250 on Build GRCh38
rs1037420753 Chr.6: 169740124-169740124 on Build GRCh38
rs373714328 Chr.6: 169740180-169740180 on Build GRCh38
rs141144989 Chr.6: 169740217-169740217 on Build GRCh38
rs779288918 Chr.6: 169740183-169740183 on Build GRCh38
rs185983441 Chr.6: 169740152-169740152 on Build GRCh38
rs778139892 Chr.6: 169740201-169740201 on Build GRCh38
rs557127049 Chr.6: 169740109-169740109 on Build GRCh38
rs755074306 Chr.6: 169740211-169740211 on Build GRCh38
rs138550412 Chr.6: 169740238-169740238 on Build GRCh38
rs772962511 Chr.6: 169740239-169740239 on Build GRCh38
rs144598809 Chr.6: 169740223-169740223 on Build GRCh38
rs372752471 Chr.6: 169740177-169740177 on Build GRCh38
rs767338730 Chr.6: 169740150-169740150 on Build GRCh38
rs773463829 Chr.6: 169740155-169740155 on Build GRCh38
rs760251746 Chr.6: 169740246-169740246 on Build GRCh38
rs369854479 Chr.6: 169740153-169740153 on Build GRCh38
rs750371781 Chr.6: 169740133-169740133 on Build GRCh38
rs188484653 Chr.6: 169740230-169740230 on Build GRCh38
rs772625449 Chr.6: 169740194-169740194 on Build GRCh38
rs989779694 Chr.6: 169740252-169740252 on Build GRCh38
rs890047333 Chr.6: 169740159-169740159 on Build GRCh38
rs897197319 Chr.6: 169740126-169740126 on Build GRCh38
rs747117883 Chr.6: 169740202-169740202 on Build GRCh38
rs766233216 Chr.6: 169740205-169740206 on Build GRCh38
rs776936731 Chr.6: 169740218-169740218 on Build GRCh38
rs779210558 Chr.6: 169740166-169740179 on Build GRCh38
rs774340519 Chr.6: 169740225-169740225 on Build GRCh38
rs757208332 Chr.6: 169740224-169740224 on Build GRCh38
rs201340121 Chr.6: 169740193-169740193 on Build GRCh38
rs199858494 Chr.6: 169740192-169740192 on Build GRCh38
rs753312915 Chr.6: 169740251-169740251 on Build GRCh38
rs750011760 Chr.6: 169740151-169740151 on Build GRCh38

View Full Product Details