Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31061154-31061411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF668
Entrez Gene ID
79759
Gene Name
zinc finger protein 668
Gene Aliases
-
UniGene
Hs.102928
Gene Chromosome Location
Chr.16: 31060843-31074320 on Build GRCh38
Transcripts
NM_001172668.1 NM_001172669.1 NM_001172670.1 NM_024706.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5107117 c.1586C>T p.T529M Chr.16: 31061342-31061342 on Build GRCh38
4471162 c.1633C>T p.P545S Chr.16: 31061295-31061295 on Build GRCh38
5954127 c.1610C>T p.S537L Chr.16: 31061318-31061318 on Build GRCh38
3377873 c.1824C>T p.S608S Chr.16: 31061173-31061173 on Build GRCh38
5420442 c.1788C>A p.T596T Chr.16: 31061209-31061209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906879708 Chr.16: 31061390-31061390 on Build GRCh38
rs778576074 Chr.16: 31061174-31061174 on Build GRCh38
rs895138371 Chr.16: 31061276-31061276 on Build GRCh38
rs371391056 Chr.16: 31061217-31061217 on Build GRCh38
rs140466221 Chr.16: 31061338-31061338 on Build GRCh38
rs778573492 Chr.16: 31061272-31061272 on Build GRCh38
rs199848754 Chr.16: 31061279-31061279 on Build GRCh38
rs771323287 Chr.16: 31061321-31061321 on Build GRCh38
rs77284397 Chr.16: 31061385-31061385 on Build GRCh38
rs767695513 Chr.16: 31061249-31061249 on Build GRCh38
rs771500094 Chr.16: 31061189-31061189 on Build GRCh38
rs757016386 Chr.16: 31061267-31061267 on Build GRCh38
rs753738967 Chr.16: 31061346-31061346 on Build GRCh38
rs745516518 Chr.16: 31061184-31061184 on Build GRCh38
rs375678762 Chr.16: 31061254-31061254 on Build GRCh38
rs976719152 Chr.16: 31061365-31061365 on Build GRCh38
rs770091266 Chr.16: 31061385-31061385 on Build GRCh38
rs757205326 Chr.16: 31061347-31061347 on Build GRCh38
rs865841504 Chr.16: 31061228-31061228 on Build GRCh38
rs760791449 Chr.16: 31061335-31061335 on Build GRCh38
rs770333162 Chr.16: 31061212-31061212 on Build GRCh38
rs376316594 Chr.16: 31061206-31061206 on Build GRCh38
rs150560360 Chr.16: 31061322-31061322 on Build GRCh38
rs377278400 Chr.16: 31061383-31061383 on Build GRCh38
rs1044428807 Chr.16: 31061352-31061352 on Build GRCh38
rs7187220 Chr.16: 31061286-31061286 on Build GRCh38
rs763376126 Chr.16: 31061320-31061320 on Build GRCh38
rs907084283 Chr.16: 31061226-31061226 on Build GRCh38
rs773712024 Chr.16: 31061311-31061311 on Build GRCh38
rs756586541 Chr.16: 31061281-31061281 on Build GRCh38
rs763330703 Chr.16: 31061220-31061220 on Build GRCh38
rs936861569 Chr.16: 31061280-31061280 on Build GRCh38
rs750266303 Chr.16: 31061362-31061362 on Build GRCh38
rs556985038 Chr.16: 31061334-31061334 on Build GRCh38
rs777728754 Chr.16: 31061262-31061262 on Build GRCh38
rs373478949 Chr.16: 31061295-31061295 on Build GRCh38
rs765063275 Chr.16: 31061360-31061360 on Build GRCh38
rs753739735 Chr.16: 31061263-31061263 on Build GRCh38
rs553391929 Chr.16: 31061234-31061234 on Build GRCh38
rs199894608 Chr.16: 31061372-31061372 on Build GRCh38
rs140247361 Chr.16: 31061299-31061299 on Build GRCh38
rs751738411 Chr.16: 31061238-31061238 on Build GRCh38
rs746680234 Chr.16: 31061391-31061391 on Build GRCh38
rs760397907 Chr.16: 31061384-31061385 on Build GRCh38
rs766668962 Chr.16: 31061229-31061229 on Build GRCh38
rs370523796 Chr.16: 31061304-31061304 on Build GRCh38
rs775048014 Chr.16: 31061194-31061194 on Build GRCh38
rs542747791 Chr.16: 31061348-31061348 on Build GRCh38
rs895428696 Chr.16: 31061307-31061307 on Build GRCh38
rs767895927 Chr.16: 31061325-31061325 on Build GRCh38
rs779640003 Chr.16: 31061389-31061389 on Build GRCh38
rs777174663 Chr.16: 31061250-31061250 on Build GRCh38
rs148738674 Chr.16: 31061261-31061261 on Build GRCh38

View Full Product Details