Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 141230205-141230460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF330
Entrez Gene ID
27309
Gene Name
zinc finger protein 330
Gene Aliases
HSA6591, NOA36
UniGene
Hs.120766
Gene Chromosome Location
Chr.4: 141220294-141234697 on Build GRCh38
Transcripts
NM_001292002.1 NM_014487.5 XM_017008033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4186381 c.460T>C p.C154R Chr.4: 141230207-141230207 on Build GRCh38
480904 c.465A>G p.E155E Chr.4: 141230212-141230212 on Build GRCh38
6332993 c.498G>T p.Q166H Chr.4: 141230245-141230245 on Build GRCh38
447421 c.477T>A p.F159L Chr.4: 141230224-141230224 on Build GRCh38
1271282 c.462T>G p.C154W Chr.4: 141230209-141230209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769734196 Chr.4: 141230250-141230250 on Build GRCh38
rs963977941 Chr.4: 141230327-141230327 on Build GRCh38
rs985096071 Chr.4: 141230284-141230284 on Build GRCh38
rs762493674 Chr.4: 141230269-141230269 on Build GRCh38
rs772116789 Chr.4: 141230299-141230300 on Build GRCh38
rs765997032 Chr.4: 141230278-141230278 on Build GRCh38
rs372351801 Chr.4: 141230309-141230309 on Build GRCh38
rs973708547 Chr.4: 141230329-141230329 on Build GRCh38
rs367614532 Chr.4: 141230295-141230295 on Build GRCh38
rs1057481093 Chr.4: 141230427-141230427 on Build GRCh38
rs373479241 Chr.4: 141230236-141230236 on Build GRCh38
rs376372235 Chr.4: 141230292-141230292 on Build GRCh38
rs929527894 Chr.4: 141230423-141230423 on Build GRCh38
rs767708538 Chr.4: 141230307-141230307 on Build GRCh38
rs772692936 Chr.4: 141230251-141230251 on Build GRCh38
rs732624 Chr.4: 141230419-141230419 on Build GRCh38

View Full Product Details