Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75070538-75070809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC2HC1C
Entrez Gene ID
79696
Gene Name
zinc finger C2HC-type containing 1C
Gene Aliases
C14orf140, FAM164C
UniGene
Hs.48642
Gene Chromosome Location
Chr.14: 75069577-75079987 on Build GRCh38
Transcripts
NM_001042430.2 NM_024643.3 NR_110315.1 XM_005268062.3 XM_005268063.4 XM_006720255.3 XM_011537155.2 XM_011537156.2 XM_011537157.2 XM_017021643.1 XM_017021644.1 XM_017021645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599319 c.153C>G p.N51K Chr.14: 75070726-75070726 on Build GRCh38
4440853 c.206A>T p.Y69F Chr.14: 75070779-75070779 on Build GRCh38
5616796 c.165G>A p.K55K Chr.14: 75070738-75070738 on Build GRCh38
7220289 c.105C>T p.Y35Y Chr.14: 75070678-75070678 on Build GRCh38
1477775 c.42C>T p.G14G Chr.14: 75070615-75070615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780135750 Chr.14: 75070629-75070629 on Build GRCh38
rs774190689 Chr.14: 75070784-75070784 on Build GRCh38
rs151153588 Chr.14: 75070618-75070619 on Build GRCh38
rs34012003 Chr.14: 75070690-75070690 on Build GRCh38
rs374941200 Chr.14: 75070675-75070675 on Build GRCh38
rs773953791 Chr.14: 75070693-75070693 on Build GRCh38
rs372846410 Chr.14: 75070583-75070583 on Build GRCh38
rs201007492 Chr.14: 75070632-75070632 on Build GRCh38
rs778393940 Chr.14: 75070669-75070669 on Build GRCh38
rs768404860 Chr.14: 75070782-75070782 on Build GRCh38
rs138703292 Chr.14: 75070711-75070711 on Build GRCh38
rs758512695 Chr.14: 75070742-75070742 on Build GRCh38
rs763751073 Chr.14: 75070596-75070596 on Build GRCh38
rs901116644 Chr.14: 75070627-75070627 on Build GRCh38
rs776559129 Chr.14: 75070754-75070755 on Build GRCh38
rs771351674 Chr.14: 75070759-75070759 on Build GRCh38
rs751125534 Chr.14: 75070597-75070597 on Build GRCh38
rs779128745 Chr.14: 75070733-75070733 on Build GRCh38
rs905642795 Chr.14: 75070577-75070577 on Build GRCh38
rs761804077 Chr.14: 75070613-75070613 on Build GRCh38
rs576142466 Chr.14: 75070682-75070682 on Build GRCh38
rs182305894 Chr.14: 75070619-75070619 on Build GRCh38
rs1000336708 Chr.14: 75070589-75070589 on Build GRCh38
rs748256536 Chr.14: 75070657-75070657 on Build GRCh38
rs192380390 Chr.14: 75070646-75070646 on Build GRCh38
rs778134011 Chr.14: 75070751-75070751 on Build GRCh38
rs190739419 Chr.14: 75070591-75070591 on Build GRCh38
rs755150388 Chr.14: 75070730-75070730 on Build GRCh38
rs772273238 Chr.14: 75070587-75070588 on Build GRCh38
rs552859440 Chr.14: 75070686-75070686 on Build GRCh38
rs141215765 Chr.14: 75070726-75070726 on Build GRCh38
rs761926692 Chr.14: 75070777-75070785 on Build GRCh38
rs747660667 Chr.14: 75070786-75070786 on Build GRCh38
rs11546525 Chr.14: 75070678-75070678 on Build GRCh38
rs752750173 Chr.14: 75070738-75070738 on Build GRCh38
rs747165560 Chr.14: 75070652-75070653 on Build GRCh38
rs747605833 Chr.14: 75070754-75070754 on Build GRCh38
rs761461296 Chr.14: 75070697-75070697 on Build GRCh38
rs767535751 Chr.14: 75070705-75070705 on Build GRCh38
rs992427245 Chr.14: 75070732-75070732 on Build GRCh38
rs746260201 Chr.14: 75070766-75070766 on Build GRCh38
rs201224540 Chr.14: 75070647-75070647 on Build GRCh38
rs768989905 Chr.14: 75070711-75070713 on Build GRCh38
rs202071124 Chr.14: 75070643-75070643 on Build GRCh38
rs762361502 Chr.14: 75070592-75070592 on Build GRCh38
rs772518985 Chr.14: 75070668-75070668 on Build GRCh38
rs746402551 Chr.14: 75070579-75070579 on Build GRCh38
rs375361819 Chr.14: 75070715-75070715 on Build GRCh38
rs187674277 Chr.14: 75070620-75070620 on Build GRCh38
rs766141700 Chr.14: 75070720-75070720 on Build GRCh38

View Full Product Details