Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15651049-15651276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM16
Entrez Gene ID
10626
Gene Name
tripartite motif containing 16
Gene Aliases
EBBP
UniGene
Hs.123534
Gene Chromosome Location
Chr.17: 15627966-15682879 on Build GRCh38
Transcripts
NM_006470.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5440041 c.338A>G p.K113R Chr.17: 15651272-15651272 on Build GRCh38
148197 c.363G>T p.E121D Chr.17: 15651247-15651247 on Build GRCh38
3765993 c.420C>T p.A140A Chr.17: 15651190-15651190 on Build GRCh38
5126878 c.449G>A p.C150Y Chr.17: 15651161-15651161 on Build GRCh38
4671277 c.505C>T p.R169C Chr.17: 15651105-15651105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145594570 Chr.17: 15651223-15651223 on Build GRCh38
rs4792642 Chr.17: 15651190-15651190 on Build GRCh38
rs754499963 Chr.17: 15651178-15651178 on Build GRCh38
rs937381211 Chr.17: 15651249-15651249 on Build GRCh38
rs528978044 Chr.17: 15651225-15651225 on Build GRCh38
rs766231260 Chr.17: 15651167-15651167 on Build GRCh38
rs1000248219 Chr.17: 15651079-15651079 on Build GRCh38
rs776009860 Chr.17: 15651108-15651108 on Build GRCh38
rs772482617 Chr.17: 15651105-15651105 on Build GRCh38
rs776896442 Chr.17: 15651125-15651125 on Build GRCh38
rs200262081 Chr.17: 15651134-15651134 on Build GRCh38
rs1026773661 Chr.17: 15651140-15651140 on Build GRCh38
rs1042011844 Chr.17: 15651069-15651069 on Build GRCh38
rs746201378 Chr.17: 15651104-15651104 on Build GRCh38
rs144568697 Chr.17: 15651191-15651191 on Build GRCh38
rs780822886 Chr.17: 15651180-15651180 on Build GRCh38
rs772418858 Chr.17: 15651206-15651206 on Build GRCh38
rs1007320809 Chr.17: 15651119-15651119 on Build GRCh38
rs751508968 Chr.17: 15651173-15651173 on Build GRCh38
rs657380 Chr.17: 15651217-15651217 on Build GRCh38
rs771467114 Chr.17: 15651088-15651088 on Build GRCh38
rs144630893 Chr.17: 15651224-15651224 on Build GRCh38
rs762722832 Chr.17: 15651252-15651252 on Build GRCh38
rs775534126 Chr.17: 15651077-15651077 on Build GRCh38
rs879000045 Chr.17: 15651154-15651155 on Build GRCh38
rs779387833 Chr.17: 15651091-15651091 on Build GRCh38
rs945420249 Chr.17: 15651245-15651245 on Build GRCh38
rs657363 Chr.17: 15651202-15651202 on Build GRCh38
rs781057869 Chr.17: 15651074-15651074 on Build GRCh38
rs373394688 Chr.17: 15651078-15651078 on Build GRCh38
rs376571595 Chr.17: 15651116-15651116 on Build GRCh38
rs201801259 Chr.17: 15651146-15651146 on Build GRCh38
rs61741220 Chr.17: 15651142-15651142 on Build GRCh38
rs562799935 Chr.17: 15651153-15651153 on Build GRCh38
rs765739065 Chr.17: 15651156-15651156 on Build GRCh38
rs762748965 Chr.17: 15651148-15651148 on Build GRCh38
rs762002659 Chr.17: 15651128-15651128 on Build GRCh38
rs560522804 Chr.17: 15651082-15651082 on Build GRCh38
rs773415380 Chr.17: 15651250-15651250 on Build GRCh38
rs2074890 Chr.17: 15651247-15651247 on Build GRCh38
rs754444064 Chr.17: 15651066-15651066 on Build GRCh38
rs776662920 Chr.17: 15651243-15651243 on Build GRCh38
rs560028798 Chr.17: 15651200-15651200 on Build GRCh38

View Full Product Details