Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 70594511-70594738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAM1
Entrez Gene ID
23471
Gene Name
translocation associated membrane protein 1
Gene Aliases
PNAS8, TRAM, TRAMP
UniGene
Hs.491988
Gene Chromosome Location
Chr.8: 70573218-70608459 on Build GRCh38
Transcripts
NM_001317804.1 NM_001317805.1 NM_014294.5 XM_017013266.1 XM_017013267.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
750795 c.493A>G p.M165V Chr.8: 70594583-70594583 on Build GRCh38
4735745 c.518T>A p.L173Q Chr.8: 70594558-70594558 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs920792581 Chr.8: 70594697-70594697 on Build GRCh38
rs185330775 Chr.8: 70594694-70594694 on Build GRCh38
rs144158109 Chr.8: 70594709-70594709 on Build GRCh38
rs759646038 Chr.8: 70594578-70594578 on Build GRCh38
rs550505259 Chr.8: 70594557-70594557 on Build GRCh38
rs774373459 Chr.8: 70594570-70594570 on Build GRCh38
rs752748180 Chr.8: 70594593-70594593 on Build GRCh38
rs767533504 Chr.8: 70594584-70594584 on Build GRCh38
rs767504568 Chr.8: 70594601-70594601 on Build GRCh38
rs545659000 Chr.8: 70594568-70594568 on Build GRCh38
rs758102006 Chr.8: 70594620-70594620 on Build GRCh38
rs755468713 Chr.8: 70594596-70594596 on Build GRCh38
rs10112383 Chr.8: 70594640-70594640 on Build GRCh38
rs778629385 Chr.8: 70594616-70594616 on Build GRCh38
rs754286488 Chr.8: 70594612-70594612 on Build GRCh38
rs776301382 Chr.8: 70594632-70594633 on Build GRCh38
rs1000880117 Chr.8: 70594646-70594646 on Build GRCh38
rs953505988 Chr.8: 70594630-70594630 on Build GRCh38
rs764500489 Chr.8: 70594609-70594609 on Build GRCh38
rs373045164 Chr.8: 70594613-70594613 on Build GRCh38
rs577864059 Chr.8: 70594693-70594693 on Build GRCh38
rs746514025 Chr.8: 70594631-70594631 on Build GRCh38
rs57799438 Chr.8: 70594639-70594639 on Build GRCh38
rs555304525 Chr.8: 70594669-70594669 on Build GRCh38
rs992187818 Chr.8: 70594710-70594710 on Build GRCh38
rs932805567 Chr.8: 70594600-70594600 on Build GRCh38
rs1009236890 Chr.8: 70594708-70594708 on Build GRCh38
rs779753143 Chr.8: 70594623-70594623 on Build GRCh38
rs759163139 Chr.8: 70594633-70594633 on Build GRCh38
rs543683519 Chr.8: 70594650-70594653 on Build GRCh38
rs750054226 Chr.8: 70594617-70594617 on Build GRCh38
rs768776356 Chr.8: 70594632-70594632 on Build GRCh38
rs774652243 Chr.8: 70594641-70594641 on Build GRCh38
rs764949026 Chr.8: 70594639-70594640 on Build GRCh38
rs575151010 Chr.8: 70594581-70594581 on Build GRCh38

View Full Product Details