Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 81336592-81336851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC3
Entrez Gene ID
342125
Gene Name
transmembrane channel like 3
Gene Aliases
-
UniGene
Hs.253489
Gene Chromosome Location
Chr.15: 81332419-81374077 on Build GRCh38
Transcripts
NM_001080532.1 NR_120365.1

Gene Symbol
TMC3-AS1
Entrez Gene ID
101929655
Gene Name
TMC3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.696654
Gene Chromosome Location
Chr.15: 81324333-81443076 on Build GRCh38
Transcripts
NM_001080532.1 NR_120365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
416790 c.2200G>C p.E734Q Chr.15: 81336612-81336612 on Build GRCh38
3401958 c.2203+2T>C p.? Chr.15: 81336607-81336607 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs543647246 Chr.15: 81336705-81336705 on Build GRCh38
rs768422580 Chr.15: 81336669-81336669 on Build GRCh38
rs778039188 Chr.15: 81336653-81336653 on Build GRCh38
rs566044606 Chr.15: 81336816-81336816 on Build GRCh38
rs943845931 Chr.15: 81336720-81336720 on Build GRCh38
rs371105295 Chr.15: 81336657-81336657 on Build GRCh38
rs779487401 Chr.15: 81336622-81336622 on Build GRCh38
rs775145242 Chr.15: 81336659-81336659 on Build GRCh38
rs766109435 Chr.15: 81336666-81336667 on Build GRCh38
rs538351687 Chr.15: 81336788-81336788 on Build GRCh38
rs552715267 Chr.15: 81336789-81336789 on Build GRCh38
rs1053508905 Chr.15: 81336796-81336796 on Build GRCh38
rs982687836 Chr.15: 81336647-81336647 on Build GRCh38
rs757289780 Chr.15: 81336793-81336793 on Build GRCh38
rs113001429 Chr.15: 81336778-81336778 on Build GRCh38
rs938759450 Chr.15: 81336673-81336673 on Build GRCh38
rs774372344 Chr.15: 81336671-81336671 on Build GRCh38
rs1031311989 Chr.15: 81336798-81336798 on Build GRCh38
rs772309132 Chr.15: 81336629-81336629 on Build GRCh38
rs894313347 Chr.15: 81336785-81336785 on Build GRCh38
rs771338206 Chr.15: 81336658-81336658 on Build GRCh38
rs150024184 Chr.15: 81336668-81336668 on Build GRCh38
rs892126128 Chr.15: 81336802-81336802 on Build GRCh38
rs748127824 Chr.15: 81336627-81336627 on Build GRCh38
rs1009197318 Chr.15: 81336812-81336812 on Build GRCh38
rs777801706 Chr.15: 81336745-81336745 on Build GRCh38

View Full Product Details