Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2807575-2807800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THOP1
Entrez Gene ID
7064
Gene Name
thimet oligopeptidase 1
Gene Aliases
EP24.15, MEPD_HUMAN, MP78, TOP
UniGene
Hs.78769
Gene Chromosome Location
Chr.19: 2785460-2813604 on Build GRCh38
Transcripts
NM_003249.4 XM_011528228.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5540343 c.1201G>A p.D401N Chr.19: 2807756-2807756 on Build GRCh38
1711599 c.1081C>T p.P361S Chr.19: 2807636-2807636 on Build GRCh38
710582 c.1204G>A p.A402T Chr.19: 2807759-2807759 on Build GRCh38
5642523 c.1120C>T p.Q374* Chr.19: 2807675-2807675 on Build GRCh38
6801328 c.1103G>T p.G368V Chr.19: 2807658-2807658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373946615 Chr.19: 2807739-2807739 on Build GRCh38
rs377533422 Chr.19: 2807749-2807749 on Build GRCh38
rs748851146 Chr.19: 2807780-2807780 on Build GRCh38
rs774811467 Chr.19: 2807624-2807624 on Build GRCh38
rs764205122 Chr.19: 2807758-2807758 on Build GRCh38
rs201540389 Chr.19: 2807751-2807751 on Build GRCh38
rs769242896 Chr.19: 2807738-2807738 on Build GRCh38
rs751679686 Chr.19: 2807596-2807596 on Build GRCh38
rs756438237 Chr.19: 2807752-2807752 on Build GRCh38
rs747637020 Chr.19: 2807735-2807735 on Build GRCh38
rs185080096 Chr.19: 2807598-2807598 on Build GRCh38
rs766045267 Chr.19: 2807659-2807659 on Build GRCh38
rs757826067 Chr.19: 2807760-2807760 on Build GRCh38
rs956693100 Chr.19: 2807636-2807636 on Build GRCh38
rs778488765 Chr.19: 2807612-2807612 on Build GRCh38
rs781534773 Chr.19: 2807696-2807696 on Build GRCh38
rs746192149 Chr.19: 2807764-2807764 on Build GRCh38
rs771375048 Chr.19: 2807620-2807620 on Build GRCh38
rs141969247 Chr.19: 2807693-2807693 on Build GRCh38
rs776679853 Chr.19: 2807629-2807629 on Build GRCh38
rs762807311 Chr.19: 2807740-2807740 on Build GRCh38
rs147544038 Chr.19: 2807595-2807595 on Build GRCh38
rs762094441 Chr.19: 2807639-2807639 on Build GRCh38
rs747755260 Chr.19: 2807774-2807774 on Build GRCh38
rs773107953 Chr.19: 2807705-2807705 on Build GRCh38
rs749774222 Chr.19: 2807713-2807713 on Build GRCh38
rs527473166 Chr.19: 2807657-2807657 on Build GRCh38
rs765389968 Chr.19: 2807648-2807648 on Build GRCh38
rs761383414 Chr.19: 2807647-2807647 on Build GRCh38
rs749637065 Chr.19: 2807617-2807617 on Build GRCh38
rs780308178 Chr.19: 2807771-2807771 on Build GRCh38
rs765131589 Chr.19: 2807652-2807652 on Build GRCh38
rs753980550 Chr.19: 2807759-2807759 on Build GRCh38
rs537514244 Chr.19: 2807719-2807719 on Build GRCh38
rs148139735 Chr.19: 2807603-2807603 on Build GRCh38
rs567921382 Chr.19: 2807761-2807761 on Build GRCh38
rs1022377947 Chr.19: 2807653-2807653 on Build GRCh38
rs200598292 Chr.19: 2807638-2807638 on Build GRCh38
rs150526087 Chr.19: 2807734-2807734 on Build GRCh38
rs777040591 Chr.19: 2807608-2807608 on Build GRCh38
rs566776905 Chr.19: 2807656-2807656 on Build GRCh38
rs766348860 Chr.19: 2807743-2807743 on Build GRCh38
rs758849669 Chr.19: 2807766-2807766 on Build GRCh38
rs755256017 Chr.19: 2807597-2807597 on Build GRCh38
rs768193342 Chr.19: 2807729-2807729 on Build GRCh38
rs370402164 Chr.19: 2807750-2807750 on Build GRCh38
rs535432531 Chr.19: 2807779-2807779 on Build GRCh38
rs752714345 Chr.19: 2807704-2807704 on Build GRCh38
rs760179127 Chr.19: 2807632-2807632 on Build GRCh38
rs570540421 Chr.19: 2807712-2807712 on Build GRCh38
rs757846063 Chr.19: 2807714-2807714 on Build GRCh38
rs940668502 Chr.19: 2807663-2807663 on Build GRCh38
rs751370013 Chr.19: 2807684-2807684 on Build GRCh38

View Full Product Details