Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3388282-3388547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC12
Entrez Gene ID
51112
Gene Name
trafficking protein particle complex 12
Gene Aliases
CGI-87, TTC-15, TTC15
UniGene
Hs.252713
Gene Chromosome Location
Chr.2: 3379245-3479571 on Build GRCh38
Transcripts
NM_001321102.1 NM_016030.5 XM_011510350.2 XM_011510352.2 XM_011510353.2 XM_011510354.2 XM_017004257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6428060 c.661T>C p.S221P Chr.2: 3388284-3388284 on Build GRCh38
5862468 c.797G>A p.R266Q Chr.2: 3388420-3388420 on Build GRCh38
4093997 c.873C>T p.D291D Chr.2: 3388496-3388496 on Build GRCh38
1230742 c.790G>C p.A264P Chr.2: 3388413-3388413 on Build GRCh38
6431498 c.764G>T p.G255V Chr.2: 3388387-3388387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374028722 Chr.2: 3388427-3388427 on Build GRCh38
rs994752260 Chr.2: 3388487-3388487 on Build GRCh38
rs1055218864 Chr.2: 3388355-3388355 on Build GRCh38
rs147425056 Chr.2: 3388472-3388472 on Build GRCh38
rs758348865 Chr.2: 3388404-3388404 on Build GRCh38
rs149516130 Chr.2: 3388433-3388433 on Build GRCh38
rs756236682 Chr.2: 3388347-3388347 on Build GRCh38
rs774857814 Chr.2: 3388423-3388423 on Build GRCh38
rs565484699 Chr.2: 3388365-3388365 on Build GRCh38
rs767405624 Chr.2: 3388451-3388451 on Build GRCh38
rs368604120 Chr.2: 3388310-3388310 on Build GRCh38
rs755724563 Chr.2: 3388457-3388457 on Build GRCh38
rs762010438 Chr.2: 3388439-3388439 on Build GRCh38
rs201138691 Chr.2: 3388518-3388518 on Build GRCh38
rs772472614 Chr.2: 3388478-3388478 on Build GRCh38
rs752446613 Chr.2: 3388453-3388453 on Build GRCh38
rs143380863 Chr.2: 3388477-3388477 on Build GRCh38
rs965772829 Chr.2: 3388514-3388514 on Build GRCh38
rs142941546 Chr.2: 3388464-3388464 on Build GRCh38
rs956986904 Chr.2: 3388527-3388527 on Build GRCh38
rs548068716 Chr.2: 3388426-3388426 on Build GRCh38
rs777042481 Chr.2: 3388502-3388502 on Build GRCh38
rs777434395 Chr.2: 3388462-3388462 on Build GRCh38
rs1015810337 Chr.2: 3388370-3388370 on Build GRCh38
rs10865541 Chr.2: 3388304-3388304 on Build GRCh38
rs767211733 Chr.2: 3388421-3388421 on Build GRCh38
rs752675378 Chr.2: 3388345-3388345 on Build GRCh38
rs906542332 Chr.2: 3388338-3388338 on Build GRCh38
rs768954505 Chr.2: 3388501-3388501 on Build GRCh38
rs753525355 Chr.2: 3388526-3388526 on Build GRCh38
rs770066925 Chr.2: 3388506-3388506 on Build GRCh38
rs774631193 Chr.2: 3388385-3388385 on Build GRCh38
rs748800063 Chr.2: 3388422-3388422 on Build GRCh38
rs751350622 Chr.2: 3388409-3388409 on Build GRCh38
rs1036286581 Chr.2: 3388466-3388466 on Build GRCh38
rs754550846 Chr.2: 3388359-3388359 on Build GRCh38
rs766454686 Chr.2: 3388407-3388407 on Build GRCh38
rs780678491 Chr.2: 3388414-3388414 on Build GRCh38
rs753982817 Chr.2: 3388402-3388402 on Build GRCh38
rs769646054 Chr.2: 3388353-3388354 on Build GRCh38
rs11686212 Chr.2: 3388524-3388524 on Build GRCh38
rs200238233 Chr.2: 3388353-3388353 on Build GRCh38
rs778690090 Chr.2: 3388351-3388351 on Build GRCh38
rs189855582 Chr.2: 3388301-3388301 on Build GRCh38
rs200381361 Chr.2: 3388356-3388356 on Build GRCh38
rs752496175 Chr.2: 3388520-3388520 on Build GRCh38
rs760638634 Chr.2: 3388399-3388399 on Build GRCh38
rs766793590 Chr.2: 3388314-3388314 on Build GRCh38
rs763275430 Chr.2: 3388313-3388313 on Build GRCh38
rs532843039 Chr.2: 3388398-3388398 on Build GRCh38
rs371953434 Chr.2: 3388324-3388324 on Build GRCh38
rs759695269 Chr.2: 3388325-3388325 on Build GRCh38
rs888937835 Chr.2: 3388492-3388492 on Build GRCh38
rs772816068 Chr.2: 3388423-3388424 on Build GRCh38
rs749851940 Chr.2: 3388377-3388377 on Build GRCh38
rs754726611 Chr.2: 3388441-3388441 on Build GRCh38
rs780402663 Chr.2: 3388490-3388490 on Build GRCh38
rs753291838 Chr.2: 3388463-3388463 on Build GRCh38
rs746808198 Chr.2: 3388358-3388358 on Build GRCh38
rs746192388 Chr.2: 3388425-3388425 on Build GRCh38
rs368230760 Chr.2: 3388521-3388521 on Build GRCh38
rs764295495 Chr.2: 3388401-3388401 on Build GRCh38
rs776815310 Chr.2: 3388438-3388438 on Build GRCh38
rs758257038 Chr.2: 3388354-3388354 on Build GRCh38
rs769386931 Chr.2: 3388374-3388374 on Build GRCh38
rs149828348 Chr.2: 3388495-3388495 on Build GRCh38
rs780920005 Chr.2: 3388360-3388360 on Build GRCh38
rs764419569 Chr.2: 3388434-3388434 on Build GRCh38
rs767359170 Chr.2: 3388515-3388515 on Build GRCh38
rs773702217 Chr.2: 3388376-3388376 on Build GRCh38
rs747590362 Chr.2: 3388372-3388372 on Build GRCh38
rs965387790 Chr.2: 3388315-3388315 on Build GRCh38
rs754852358 Chr.2: 3388410-3388410 on Build GRCh38
rs200068871 Chr.2: 3388529-3388529 on Build GRCh38
rs998158367 Chr.2: 3388522-3388522 on Build GRCh38
rs371542203 Chr.2: 3388417-3388417 on Build GRCh38
rs759458791 Chr.2: 3388512-3388512 on Build GRCh38
rs866676342 Chr.2: 3388496-3388496 on Build GRCh38
rs749106989 Chr.2: 3388453-3388454 on Build GRCh38
rs199789871 Chr.2: 3388440-3388440 on Build GRCh38
rs747903695 Chr.2: 3388416-3388416 on Build GRCh38
rs531841765 Chr.2: 3388337-3388337 on Build GRCh38
rs767583645 Chr.2: 3388392-3388392 on Build GRCh38
rs773820162 Chr.2: 3388312-3388312 on Build GRCh38
rs367769506 Chr.2: 3388386-3388386 on Build GRCh38
rs774144539 Chr.2: 3388511-3388511 on Build GRCh38
rs146562015 Chr.2: 3388303-3388303 on Build GRCh38
rs779358370 Chr.2: 3388473-3388473 on Build GRCh38

View Full Product Details