Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34855548-34855805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM12
Entrez Gene ID
113444
Gene Name
small integral membrane protein 12
Gene Aliases
C1orf212
UniGene
Hs.27160
Gene Chromosome Location
Chr.1: 34850362-34859820 on Build GRCh38
Transcripts
NM_001164824.1 NM_001164825.1 NM_001320261.1 NM_138428.5 XM_005270403.4 XM_011540553.2 XM_011540554.2 XM_011540555.2 XM_017000166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4532479 c.188G>A p.G63D Chr.1: 34855790-34855790 on Build GRCh38
5992228 c.108G>T p.Q36H Chr.1: 34855573-34855573 on Build GRCh38
1602265 c.1A>G p.M1V Chr.1: 34855680-34855680 on Build GRCh38
908588 c.12G>A p.L4L Chr.1: 34855669-34855669 on Build GRCh38
908587 c.55A>C p.N19H Chr.1: 34855626-34855626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898164579 Chr.1: 34855773-34855773 on Build GRCh38
rs759754866 Chr.1: 34855778-34855778 on Build GRCh38
rs80184073 Chr.1: 34855690-34855690 on Build GRCh38
rs748997209 Chr.1: 34855619-34855619 on Build GRCh38
rs542025856 Chr.1: 34855575-34855575 on Build GRCh38
rs933860429 Chr.1: 34855578-34855578 on Build GRCh38
rs781581623 Chr.1: 34855680-34855680 on Build GRCh38
rs748600537 Chr.1: 34855583-34855583 on Build GRCh38
rs763230792 Chr.1: 34855663-34855663 on Build GRCh38
rs755304095 Chr.1: 34855677-34855677 on Build GRCh38
rs981615058 Chr.1: 34855745-34855745 on Build GRCh38
rs370089432 Chr.1: 34855689-34855689 on Build GRCh38
rs755567962 Chr.1: 34855571-34855571 on Build GRCh38
rs145113026 Chr.1: 34855671-34855671 on Build GRCh38
rs777465466 Chr.1: 34855691-34855691 on Build GRCh38
rs745455058 Chr.1: 34855738-34855738 on Build GRCh38
rs774468275 Chr.1: 34855777-34855777 on Build GRCh38
rs376951799 Chr.1: 34855766-34855766 on Build GRCh38
rs768580691 Chr.1: 34855743-34855743 on Build GRCh38
rs753993701 Chr.1: 34855715-34855715 on Build GRCh38
rs771790970 Chr.1: 34855739-34855739 on Build GRCh38
rs746992765 Chr.1: 34855742-34855742 on Build GRCh38
rs750474172 Chr.1: 34855655-34855655 on Build GRCh38
rs899636611 Chr.1: 34855570-34855570 on Build GRCh38
rs935540185 Chr.1: 34855734-34855734 on Build GRCh38
rs759236495 Chr.1: 34855631-34855631 on Build GRCh38
rs774268090 Chr.1: 34855623-34855623 on Build GRCh38
rs14103 Chr.1: 34855641-34855641 on Build GRCh38
rs762958112 Chr.1: 34855761-34855761 on Build GRCh38
rs528788618 Chr.1: 34855741-34855741 on Build GRCh38
rs770458181 Chr.1: 34855620-34855620 on Build GRCh38
rs775406423 Chr.1: 34855643-34855643 on Build GRCh38
rs138225599 Chr.1: 34855650-34855650 on Build GRCh38
rs770673301 Chr.1: 34855729-34855729 on Build GRCh38
rs778440742 Chr.1: 34855735-34855735 on Build GRCh38
rs777369856 Chr.1: 34855618-34855618 on Build GRCh38
rs760561697 Chr.1: 34855644-34855644 on Build GRCh38
rs138624697 Chr.1: 34855670-34855670 on Build GRCh38
rs769461115 Chr.1: 34855589-34855589 on Build GRCh38
rs773340373 Chr.1: 34855747-34855747 on Build GRCh38

View Full Product Details