Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184235254-184235499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA5B2
Entrez Gene ID
90113
Gene Name
von Willebrand factor A domain containing 5B2
Gene Aliases
-
UniGene
Hs.584997
Gene Chromosome Location
Chr.3: 184229593-184242329 on Build GRCh38
Transcripts
NM_001320373.1 NM_138345.2 XM_011513288.2 XM_011513289.2 XM_011513290.2 XM_011513291.2 XM_011513292.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7420579 c.1055C>T p.T352I Chr.3: 184235262-184235262 on Build GRCh38
5398751 c.1078G>A p.D360N Chr.3: 184235285-184235285 on Build GRCh38
3724156 c.1063C>T p.L355L Chr.3: 184235270-184235270 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764086572 Chr.3: 184235312-184235312 on Build GRCh38
rs535441242 Chr.3: 184235443-184235443 on Build GRCh38
rs989967884 Chr.3: 184235345-184235345 on Build GRCh38
rs553631059 Chr.3: 184235283-184235283 on Build GRCh38
rs574990447 Chr.3: 184235296-184235296 on Build GRCh38
rs557126109 Chr.3: 184235380-184235380 on Build GRCh38
rs985390699 Chr.3: 184235371-184235371 on Build GRCh38
rs923260838 Chr.3: 184235364-184235364 on Build GRCh38
rs953537071 Chr.3: 184235370-184235370 on Build GRCh38
rs575370337 Chr.3: 184235410-184235410 on Build GRCh38
rs757187000 Chr.3: 184235350-184235350 on Build GRCh38
rs192845982 Chr.3: 184235479-184235479 on Build GRCh38
rs750905791 Chr.3: 184235472-184235472 on Build GRCh38
rs543361579 Chr.3: 184235453-184235453 on Build GRCh38
rs941170009 Chr.3: 184235420-184235420 on Build GRCh38
rs879644953 Chr.3: 184235424-184235424 on Build GRCh38
rs542055716 Chr.3: 184235336-184235336 on Build GRCh38
rs964765421 Chr.3: 184235333-184235333 on Build GRCh38
rs1031015431 Chr.3: 184235297-184235297 on Build GRCh38
rs1017546542 Chr.3: 184235313-184235313 on Build GRCh38
rs976159780 Chr.3: 184235363-184235363 on Build GRCh38
rs753800076 Chr.3: 184235341-184235341 on Build GRCh38
rs780165659 Chr.3: 184235354-184235354 on Build GRCh38
rs878934363 Chr.3: 184235334-184235334 on Build GRCh38
rs911131373 Chr.3: 184235374-184235374 on Build GRCh38

View Full Product Details