Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23620491-23620724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRICH1
Entrez Gene ID
51233
Gene Name
aspartate rich 1
Gene Aliases
C22orf43, KB-208E9.1
UniGene
Hs.517466
Gene Chromosome Location
Chr.22: 23580872-23637732 on Build GRCh38
Transcripts
NM_016449.3 XM_011530206.1 XM_011530207.2 XM_011530208.2 XM_011530209.2 XM_011530210.2 XM_017028813.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3849609 c.400G>A p.V134I Chr.22: 23620600-23620600 on Build GRCh38
1415140 c.396A>G p.S132S Chr.22: 23620604-23620604 on Build GRCh38
3552538 c.391C>T p.P131S Chr.22: 23620609-23620609 on Build GRCh38
726180 c.395C>A p.S132* Chr.22: 23620605-23620605 on Build GRCh38
3964137 c.402C>A p.V134V Chr.22: 23620598-23620598 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs531569149 Chr.22: 23620608-23620608 on Build GRCh38
rs772572246 Chr.22: 23620641-23620641 on Build GRCh38
rs774597319 Chr.22: 23620627-23620627 on Build GRCh38
rs112623999 Chr.22: 23620534-23620534 on Build GRCh38
rs114580984 Chr.22: 23620684-23620684 on Build GRCh38
rs762255316 Chr.22: 23620567-23620567 on Build GRCh38
rs772777398 Chr.22: 23620598-23620598 on Build GRCh38
rs548539191 Chr.22: 23620664-23620664 on Build GRCh38
rs776177405 Chr.22: 23620651-23620651 on Build GRCh38
rs374692477 Chr.22: 23620623-23620623 on Build GRCh38
rs759144377 Chr.22: 23620652-23620652 on Build GRCh38
rs758204027 Chr.22: 23620666-23620666 on Build GRCh38
rs761156285 Chr.22: 23620643-23620643 on Build GRCh38
rs770068836 Chr.22: 23620610-23620610 on Build GRCh38
rs532804712 Chr.22: 23620604-23620604 on Build GRCh38
rs750604162 Chr.22: 23620665-23620665 on Build GRCh38
rs780171971 Chr.22: 23620616-23620616 on Build GRCh38
rs749944893 Chr.22: 23620589-23620589 on Build GRCh38
rs986613185 Chr.22: 23620613-23620613 on Build GRCh38
rs185150904 Chr.22: 23620622-23620622 on Build GRCh38
rs560661830 Chr.22: 23620603-23620603 on Build GRCh38
rs531732256 Chr.22: 23620663-23620663 on Build GRCh38
rs760351663 Chr.22: 23620682-23620682 on Build GRCh38
rs72559613 Chr.22: 23620557-23620558 on Build GRCh38
rs777798897 Chr.22: 23620607-23620607 on Build GRCh38
rs774743854 Chr.22: 23620558-23620558 on Build GRCh38
rs767963809 Chr.22: 23620570-23620573 on Build GRCh38
rs200686453 Chr.22: 23620555-23620556 on Build GRCh38
rs35192883 Chr.22: 23620592-23620593 on Build GRCh38
rs768962228 Chr.22: 23620555-23620555 on Build GRCh38
rs764895072 Chr.22: 23620661-23620661 on Build GRCh38
rs1031153550 Chr.22: 23620606-23620606 on Build GRCh38
rs765921646 Chr.22: 23620649-23620649 on Build GRCh38
rs753489168 Chr.22: 23620601-23620601 on Build GRCh38
rs369617687 Chr.22: 23620584-23620584 on Build GRCh38
rs75567718 Chr.22: 23620602-23620602 on Build GRCh38
rs773775562 Chr.22: 23620642-23620642 on Build GRCh38
rs527570783 Chr.22: 23620687-23620687 on Build GRCh38
rs767958999 Chr.22: 23620571-23620571 on Build GRCh38
rs760110073 Chr.22: 23620591-23620591 on Build GRCh38
rs748450502 Chr.22: 23620634-23620634 on Build GRCh38
rs930620630 Chr.22: 23620678-23620678 on Build GRCh38

View Full Product Details