Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4937530-4937801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDYL
Entrez Gene ID
9425
Gene Name
chromodomain protein, Y-like
Gene Aliases
CDYL1
UniGene
Hs.269092
Gene Chromosome Location
Chr.6: 4706159-4955544 on Build GRCh38
Transcripts
NM_001143970.1 NM_001143971.1 NM_004824.3 NR_026590.1 XM_011514997.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1080053 c.1192G>A p.V398I Chr.6: 4937646-4937646 on Build GRCh38
3874622 c.1259G>T p.S420I Chr.6: 4937713-4937713 on Build GRCh38
1444987 c.1179C>T p.S393S Chr.6: 4937633-4937633 on Build GRCh38
3628243 c.1136T>C p.L379P Chr.6: 4937590-4937590 on Build GRCh38
5751081 c.1153G>A p.D385N Chr.6: 4937607-4937607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779362976 Chr.6: 4937597-4937597 on Build GRCh38
rs754478809 Chr.6: 4937726-4937726 on Build GRCh38
rs750049821 Chr.6: 4937591-4937591 on Build GRCh38
rs372880737 Chr.6: 4937691-4937691 on Build GRCh38
rs200526998 Chr.6: 4937695-4937695 on Build GRCh38
rs765086192 Chr.6: 4937656-4937656 on Build GRCh38
rs779127020 Chr.6: 4937627-4937627 on Build GRCh38
rs991185648 Chr.6: 4937679-4937679 on Build GRCh38
rs368234357 Chr.6: 4937600-4937600 on Build GRCh38
rs140138635 Chr.6: 4937636-4937636 on Build GRCh38
rs149831569 Chr.6: 4937711-4937711 on Build GRCh38
rs972321836 Chr.6: 4937742-4937742 on Build GRCh38
rs200171578 Chr.6: 4937712-4937712 on Build GRCh38
rs778247904 Chr.6: 4937601-4937601 on Build GRCh38
rs141394848 Chr.6: 4937619-4937619 on Build GRCh38
rs764631579 Chr.6: 4937741-4937741 on Build GRCh38
rs147295406 Chr.6: 4937645-4937645 on Build GRCh38
rs779178574 Chr.6: 4937751-4937751 on Build GRCh38
rs761722419 Chr.6: 4937646-4937646 on Build GRCh38
rs148330526 Chr.6: 4937596-4937596 on Build GRCh38
rs376963465 Chr.6: 4937748-4937748 on Build GRCh38
rs145710209 Chr.6: 4937637-4937637 on Build GRCh38
rs751096522 Chr.6: 4937716-4937716 on Build GRCh38
rs150826447 Chr.6: 4937634-4937634 on Build GRCh38
rs539443011 Chr.6: 4937606-4937606 on Build GRCh38
rs746232500 Chr.6: 4937758-4937758 on Build GRCh38
rs754290199 Chr.6: 4937750-4937750 on Build GRCh38
rs140950492 Chr.6: 4937685-4937685 on Build GRCh38
rs749689739 Chr.6: 4937612-4937612 on Build GRCh38

View Full Product Details