Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59389281-59389552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YPEL2
Entrez Gene ID
388403
Gene Name
yippee like 2
Gene Aliases
FKSG4
UniGene
Hs.463613
Gene Chromosome Location
Chr.17: 59331692-59401734 on Build GRCh38
Transcripts
NM_001005404.3 XM_017024621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1384907 c.162-1G>C p.? Chr.17: 59389359-59389359 on Build GRCh38
6312873 c.259G>T p.G87C Chr.17: 59389457-59389457 on Build GRCh38
5055642 c.222C>T p.V74V Chr.17: 59389420-59389420 on Build GRCh38
3196327 c.184G>A p.A62T Chr.17: 59389382-59389382 on Build GRCh38
4741885 c.217G>A p.A73T Chr.17: 59389415-59389415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544344915 Chr.17: 59389356-59389356 on Build GRCh38
rs781406094 Chr.17: 59389490-59389490 on Build GRCh38
rs369765495 Chr.17: 59389516-59389516 on Build GRCh38
rs763462925 Chr.17: 59389338-59389338 on Build GRCh38
rs752924635 Chr.17: 59389472-59389481 on Build GRCh38
rs748240733 Chr.17: 59389494-59389494 on Build GRCh38
rs200204114 Chr.17: 59389339-59389341 on Build GRCh38
rs200700318 Chr.17: 59389468-59389468 on Build GRCh38
rs1026103958 Chr.17: 59389513-59389513 on Build GRCh38
rs756789747 Chr.17: 59389378-59389378 on Build GRCh38
rs114373697 Chr.17: 59389429-59389429 on Build GRCh38
rs978385343 Chr.17: 59389504-59389504 on Build GRCh38
rs766820334 Chr.17: 59389348-59389348 on Build GRCh38
rs968372464 Chr.17: 59389485-59389485 on Build GRCh38
rs773843697 Chr.17: 59389333-59389333 on Build GRCh38
rs765142424 Chr.17: 59389379-59389379 on Build GRCh38
rs186477591 Chr.17: 59389421-59389421 on Build GRCh38
rs866593262 Chr.17: 59389489-59389489 on Build GRCh38
rs564149261 Chr.17: 59389352-59389352 on Build GRCh38
rs758225787 Chr.17: 59389420-59389420 on Build GRCh38
rs748198769 Chr.17: 59389313-59389313 on Build GRCh38
rs138086928 Chr.17: 59389432-59389432 on Build GRCh38
rs770205002 Chr.17: 59389317-59389317 on Build GRCh38
rs957195150 Chr.17: 59389387-59389387 on Build GRCh38
rs760804544 Chr.17: 59389494-59389495 on Build GRCh38
rs774816814 Chr.17: 59389350-59389350 on Build GRCh38
rs763749672 Chr.17: 59389353-59389353 on Build GRCh38
rs750335791 Chr.17: 59389396-59389396 on Build GRCh38

View Full Product Details