Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69180777-69181032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4B
Entrez Gene ID
23150
Gene Name
FERM domain containing 4B
Gene Aliases
6030440G05Rik, GRSP1
UniGene
Hs.371681
Gene Chromosome Location
Chr.3: 69168782-69541340 on Build GRCh38
Transcripts
NM_015123.2 XM_005264722.1 XM_005264723.1 XM_017005989.1 XM_017005990.1 XM_017005991.1 XM_017005992.1 XM_017005993.1 XM_017005994.1 XM_017005995.1 XM_017005996.1 XM_017005997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5906290 c.2572C>T p.Q858* Chr.3: 69181016-69181016 on Build GRCh38
4415326 c.2840C>T p.S947L Chr.3: 69180910-69180910 on Build GRCh38
1137098 c.2798C>T p.A933V Chr.3: 69180952-69180952 on Build GRCh38
6666962 c.2645A>G p.Q882R Chr.3: 69180943-69180943 on Build GRCh38
7133386 c.2619G>C p.Q873H Chr.3: 69180969-69180969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970718105 Chr.3: 69181009-69181009 on Build GRCh38
rs763908685 Chr.3: 69180979-69180980 on Build GRCh38
rs372002481 Chr.3: 69180947-69180947 on Build GRCh38
rs879387393 Chr.3: 69180937-69180937 on Build GRCh38
rs758774503 Chr.3: 69180952-69180952 on Build GRCh38
rs752576449 Chr.3: 69180856-69180856 on Build GRCh38
rs533569256 Chr.3: 69180872-69180872 on Build GRCh38
rs777490803 Chr.3: 69180865-69180865 on Build GRCh38
rs760224201 Chr.3: 69180997-69180997 on Build GRCh38
rs896847888 Chr.3: 69180809-69180809 on Build GRCh38
rs779137900 Chr.3: 69180949-69180949 on Build GRCh38
rs9818314 Chr.3: 69180875-69180875 on Build GRCh38
rs143019509 Chr.3: 69180877-69180877 on Build GRCh38
rs61737530 Chr.3: 69181011-69181011 on Build GRCh38
rs561278627 Chr.3: 69180951-69180951 on Build GRCh38
rs984004857 Chr.3: 69180811-69180811 on Build GRCh38
rs1001263500 Chr.3: 69181013-69181013 on Build GRCh38
rs545542453 Chr.3: 69180878-69180878 on Build GRCh38
rs187901513 Chr.3: 69180920-69180920 on Build GRCh38
rs768563202 Chr.3: 69180976-69180976 on Build GRCh38
rs775458758 Chr.3: 69180880-69180880 on Build GRCh38
rs9831516 Chr.3: 69180910-69180910 on Build GRCh38
rs530066916 Chr.3: 69180980-69180980 on Build GRCh38
rs375403672 Chr.3: 69180803-69180803 on Build GRCh38
rs995179810 Chr.3: 69180817-69180817 on Build GRCh38
rs12054011 Chr.3: 69180825-69180825 on Build GRCh38
rs375599800 Chr.3: 69180994-69180994 on Build GRCh38
rs747045050 Chr.3: 69180958-69180958 on Build GRCh38
rs763959803 Chr.3: 69180906-69180906 on Build GRCh38
rs757455228 Chr.3: 69181012-69181012 on Build GRCh38
rs371975100 Chr.3: 69180869-69180869 on Build GRCh38
rs1012523792 Chr.3: 69180944-69180944 on Build GRCh38
rs760393219 Chr.3: 69180856-69180857 on Build GRCh38
rs574359033 Chr.3: 69180852-69180852 on Build GRCh38
rs778094939 Chr.3: 69181004-69181004 on Build GRCh38
rs199933053 Chr.3: 69180919-69180919 on Build GRCh38
rs755371188 Chr.3: 69180948-69180948 on Build GRCh38
rs750160825 Chr.3: 69180853-69180853 on Build GRCh38
rs774007665 Chr.3: 69180977-69180977 on Build GRCh38
rs902972396 Chr.3: 69180896-69180896 on Build GRCh38
rs867213130 Chr.3: 69180917-69180917 on Build GRCh38
rs780413354 Chr.3: 69180855-69180855 on Build GRCh38
rs753553130 Chr.3: 69181001-69181001 on Build GRCh38
rs540273782 Chr.3: 69181003-69181003 on Build GRCh38
rs761825533 Chr.3: 69180979-69180979 on Build GRCh38
rs951285443 Chr.3: 69180799-69180799 on Build GRCh38
rs777911678 Chr.3: 69180955-69180955 on Build GRCh38
rs13084205 Chr.3: 69180823-69180823 on Build GRCh38
rs370212737 Chr.3: 69181002-69181002 on Build GRCh38
rs1027152663 Chr.3: 69180821-69180821 on Build GRCh38
rs144393195 Chr.3: 69180918-69180918 on Build GRCh38
rs756053798 Chr.3: 69180857-69180857 on Build GRCh38
rs909789385 Chr.3: 69180815-69180815 on Build GRCh38
rs777044557 Chr.3: 69180963-69180963 on Build GRCh38
rs796959291 Chr.3: 69180875-69180877 on Build GRCh38
rs373094835 Chr.3: 69180904-69180904 on Build GRCh38
rs746057069 Chr.3: 69180974-69180974 on Build GRCh38
rs771301646 Chr.3: 69180960-69180960 on Build GRCh38

View Full Product Details