Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57753216-57753449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNB1
Entrez Gene ID
10300
Gene Name
katanin regulatory subunit B1
Gene Aliases
KAT, LIS6
UniGene
Hs.275675
Gene Chromosome Location
Chr.16: 57735717-57757250 on Build GRCh38
Transcripts
NM_005886.2 XM_005255772.4 XM_006721121.3 XM_006721122.2 XM_006721123.3 XM_011522810.2 XM_017022860.1 XM_017022861.1 XM_017022862.1 XM_017022863.1 XM_017022864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5028145 c.1001G>A p.R334Q Chr.16: 57753222-57753222 on Build GRCh38
6690284 c.1088G>C p.S363T Chr.16: 57753430-57753430 on Build GRCh38
5084750 c.1068C>T p.S356S Chr.16: 57753410-57753410 on Build GRCh38
5128541 c.995C>A p.P332H Chr.16: 57753216-57753216 on Build GRCh38
7193454 c.1010A>G p.Y337C Chr.16: 57753231-57753231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112994876 Chr.16: 57753430-57753430 on Build GRCh38
rs372138195 Chr.16: 57753236-57753236 on Build GRCh38
rs782339190 Chr.16: 57753263-57753263 on Build GRCh38
rs782445055 Chr.16: 57753238-57753238 on Build GRCh38
rs372112294 Chr.16: 57753276-57753276 on Build GRCh38
rs7185990 Chr.16: 57753235-57753235 on Build GRCh38
rs1039768697 Chr.16: 57753326-57753326 on Build GRCh38
rs377544392 Chr.16: 57753327-57753327 on Build GRCh38
rs373854173 Chr.16: 57753386-57753386 on Build GRCh38
rs555141717 Chr.16: 57753325-57753325 on Build GRCh38
rs201462999 Chr.16: 57753380-57753380 on Build GRCh38
rs781897761 Chr.16: 57753419-57753419 on Build GRCh38
rs782540305 Chr.16: 57753428-57753428 on Build GRCh38
rs376110213 Chr.16: 57753348-57753348 on Build GRCh38
rs782060401 Chr.16: 57753417-57753417 on Build GRCh38
rs782686368 Chr.16: 57753243-57753243 on Build GRCh38
rs781855687 Chr.16: 57753315-57753315 on Build GRCh38
rs782011857 Chr.16: 57753282-57753282 on Build GRCh38
rs371014561 Chr.16: 57753370-57753370 on Build GRCh38
rs781906001 Chr.16: 57753292-57753292 on Build GRCh38
rs781851855 Chr.16: 57753432-57753432 on Build GRCh38
rs934174114 Chr.16: 57753335-57753335 on Build GRCh38
rs782749135 Chr.16: 57753346-57753346 on Build GRCh38
rs142219691 Chr.16: 57753418-57753418 on Build GRCh38
rs782126778 Chr.16: 57753283-57753283 on Build GRCh38
rs782224310 Chr.16: 57753254-57753254 on Build GRCh38
rs782628714 Chr.16: 57753373-57753373 on Build GRCh38
rs782522839 Chr.16: 57753252-57753252 on Build GRCh38
rs782361800 Chr.16: 57753284-57753284 on Build GRCh38
rs782453556 Chr.16: 57753345-57753345 on Build GRCh38
rs782221901 Chr.16: 57753377-57753377 on Build GRCh38
rs782414909 Chr.16: 57753279-57753279 on Build GRCh38
rs781801088 Chr.16: 57753354-57753354 on Build GRCh38
rs572708547 Chr.16: 57753272-57753272 on Build GRCh38
rs748243417 Chr.16: 57753364-57753364 on Build GRCh38
rs200545074 Chr.16: 57753410-57753410 on Build GRCh38
rs782153019 Chr.16: 57753298-57753298 on Build GRCh38
rs373054102 Chr.16: 57753289-57753289 on Build GRCh38
rs369638304 Chr.16: 57753244-57753244 on Build GRCh38
rs369286431 Chr.16: 57753369-57753369 on Build GRCh38
rs782120101 Chr.16: 57753411-57753411 on Build GRCh38
rs782780856 Chr.16: 57753314-57753314 on Build GRCh38
rs981917778 Chr.16: 57753246-57753246 on Build GRCh38
rs782294121 Chr.16: 57753382-57753382 on Build GRCh38
rs562336850 Chr.16: 57753381-57753381 on Build GRCh38
rs782686930 Chr.16: 57753361-57753361 on Build GRCh38
rs969212898 Chr.16: 57753240-57753240 on Build GRCh38
rs782764043 Chr.16: 57753431-57753431 on Build GRCh38
rs1039017522 Chr.16: 57753392-57753392 on Build GRCh38
rs782069793 Chr.16: 57753288-57753288 on Build GRCh38
rs782429018 Chr.16: 57753358-57753358 on Build GRCh38

View Full Product Details