Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 76429189-76429432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEE1
Entrez Gene ID
57692
Gene Name
MAGE family member E1
Gene Aliases
DAMAGE, HCA1
UniGene
Hs.8453
Gene Chromosome Location
Chr.X: 76427653-76431353 on Build GRCh38
Transcripts
NM_020932.2

SNP Detail

SNP ID Chromosome Location
rs782717046 Chr.X: 76429299-76429299 on Build GRCh38
rs782556507 Chr.X: 76429339-76429339 on Build GRCh38
rs782578239 Chr.X: 76429356-76429356 on Build GRCh38
rs782808019 Chr.X: 76429210-76429210 on Build GRCh38
rs186118079 Chr.X: 76429338-76429338 on Build GRCh38
rs782208442 Chr.X: 76429349-76429349 on Build GRCh38
rs782750703 Chr.X: 76429237-76429237 on Build GRCh38
rs782637952 Chr.X: 76429341-76429341 on Build GRCh38
rs781940491 Chr.X: 76429391-76429391 on Build GRCh38
rs782152138 Chr.X: 76429395-76429395 on Build GRCh38
rs781956239 Chr.X: 76429385-76429385 on Build GRCh38
rs143682541 Chr.X: 76429390-76429390 on Build GRCh38
rs782243866 Chr.X: 76429366-76429366 on Build GRCh38
rs1004099932 Chr.X: 76429266-76429266 on Build GRCh38
rs781982088 Chr.X: 76429216-76429216 on Build GRCh38
rs781842983 Chr.X: 76429247-76429247 on Build GRCh38
rs782031633 Chr.X: 76429362-76429362 on Build GRCh38
rs782397816 Chr.X: 76429369-76429369 on Build GRCh38
rs1030363216 Chr.X: 76429403-76429403 on Build GRCh38
rs782444754 Chr.X: 76429256-76429256 on Build GRCh38
rs200401403 Chr.X: 76429351-76429351 on Build GRCh38
rs782402354 Chr.X: 76429359-76429359 on Build GRCh38
rs782132084 Chr.X: 76429235-76429235 on Build GRCh38
rs782104033 Chr.X: 76429387-76429387 on Build GRCh38

View Full Product Details