Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61867444-61867705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS2
Entrez Gene ID
57508
Gene Name
integrator complex subunit 2
Gene Aliases
INT2, KIAA1287
UniGene
Hs.279646
Gene Chromosome Location
Chr.17: 61865367-61928029 on Build GRCh38
Transcripts
NM_020748.2 NR_026641.1 XM_006721987.3 XM_011525058.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5816341 c.3599G>C p.S1200T Chr.17: 61867573-61867573 on Build GRCh38
6686412 c.3474T>A p.S1158R Chr.17: 61867698-61867698 on Build GRCh38
1385026 c.3500C>A p.S1167* Chr.17: 61867672-61867672 on Build GRCh38
3890102 c.3541C>T p.P1181S Chr.17: 61867631-61867631 on Build GRCh38
5771256 c.3484C>G p.Q1162E Chr.17: 61867688-61867688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748974026 Chr.17: 61867630-61867630 on Build GRCh38
rs201429545 Chr.17: 61867565-61867565 on Build GRCh38
rs775119135 Chr.17: 61867673-61867673 on Build GRCh38
rs770777515 Chr.17: 61867645-61867645 on Build GRCh38
rs749168057 Chr.17: 61867511-61867511 on Build GRCh38
rs745832171 Chr.17: 61867665-61867665 on Build GRCh38
rs375995084 Chr.17: 61867610-61867610 on Build GRCh38
rs760317635 Chr.17: 61867676-61867676 on Build GRCh38
rs768187449 Chr.17: 61867677-61867677 on Build GRCh38
rs199798693 Chr.17: 61867548-61867548 on Build GRCh38
rs754137778 Chr.17: 61867556-61867556 on Build GRCh38
rs772178307 Chr.17: 61867666-61867666 on Build GRCh38
rs541329984 Chr.17: 61867619-61867619 on Build GRCh38
rs372136900 Chr.17: 61867499-61867499 on Build GRCh38
rs373259898 Chr.17: 61867533-61867533 on Build GRCh38
rs767157867 Chr.17: 61867528-61867528 on Build GRCh38
rs541939682 Chr.17: 61867524-61867524 on Build GRCh38
rs571706283 Chr.17: 61867540-61867551 on Build GRCh38
rs774354018 Chr.17: 61867527-61867527 on Build GRCh38
rs758420399 Chr.17: 61867575-61867575 on Build GRCh38
rs775245724 Chr.17: 61867529-61867529 on Build GRCh38
rs137974612 Chr.17: 61867539-61867539 on Build GRCh38
rs775459191 Chr.17: 61867474-61867474 on Build GRCh38
rs1051585514 Chr.17: 61867506-61867506 on Build GRCh38
rs755182912 Chr.17: 61867613-61867613 on Build GRCh38
rs765595177 Chr.17: 61867568-61867568 on Build GRCh38
rs892535572 Chr.17: 61867517-61867517 on Build GRCh38
rs572625171 Chr.17: 61867573-61867573 on Build GRCh38
rs868392588 Chr.17: 61867498-61867498 on Build GRCh38
rs968285870 Chr.17: 61867672-61867672 on Build GRCh38
rs780136499 Chr.17: 61867609-61867609 on Build GRCh38
rs778667549 Chr.17: 61867660-61867660 on Build GRCh38

View Full Product Details